Merck
  • The ARL2 GTPase is required for mitochondrial morphology, motility, and maintenance of ATP levels.
The ARL2 GTPase is required for mitochondrial morphology, motility, and maintenance of ATP levels.
PloS one (2014-06-10)
Laura E Newman, Cheng-jing Zhou, Samatha Mudigonda, Alexa L Mattheyses, Eleonora Paradies, Carlo Marya Thomas Marobbio, Richard A Kahn
ABSTRACT
ARF-like 2 (ARL2) is a member of the ARF family and RAS superfamily of regulatory GTPases, predicted to be present in the last eukaryotic common ancestor, and essential in a number of model genetic systems. Though best studied as a regulator of tubulin folding, we previously demonstrated that ARL2 partially localizes to mitochondria. Here, we show that ARL2 is essential to a number of mitochondrial functions, including mitochondrial morphology, motility, and maintenance of ATP levels. We compare phenotypes resulting from ARL2 depletion and expression of dominant negative mutants and use these to demonstrate that the mitochondrial roles of ARL2 are distinct from its roles in tubulin folding. Testing of current models for ARL2 actions at mitochondria failed to support them. Rather, we found that knockdown of the ARL2 GTPase activating protein (GAP) ELMOD2 phenocopies two of three phenotypes of ARL2 siRNA, making it a likely effector for these actions. These results add new layers of complexity to ARL2 signaling, highlighting the need to deconvolve these different cell functions. We hypothesize that ARL2 plays essential roles inside mitochondria along with other cellular functions, at least in part to provide coupling of regulation between these essential cell processes.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Arl2
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Arl2bp
Sigma-Aldrich
MISSION® esiRNA, targeting human ARL2BP
Sigma-Aldrich
MISSION® esiRNA, targeting human ELMOD2
Sigma-Aldrich
MISSION® esiRNA, targeting human BSND
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Elmod2