Merck
  • The role of receptor for advanced glycation end products in airway inflammation in CF and CF related diabetes.
The role of receptor for advanced glycation end products in airway inflammation in CF and CF related diabetes.
Scientific reports (2015-03-11)
Siobhain Mulrennan, Svetlana Baltic, Shashi Aggarwal, Jamie Wood, Alina Miranda, Felicity Frost, Joey Kaye, Philip J Thompson
ABSTRACT
Cystic Fibrosis (CF) is often accompanied by diabetes leading to worsening lung function, the reason for which is unclear. The receptor for advanced-glycation-end-products (RAGE) regulates immune responses and inflammation and has been linked to diabetes and possibly CF. We performed a pilot study to determine if CF and CF-related diabetes (CFRD) are associated with enhanced RAGE expression. Full length (fl)RAGE, soluble (s)RAGE, endogenous soluble (es)RAGE, S100A12 (enRAGE) and advanced-glycation-end-products (AGE) expression was assessed in serum, white blood cells and sputum of patients with CF; diabetes; CFRD and healthy subjects. Sputum enRAGE/sRAGE ratios were high in CF but particularly in CFRD which negatively correlated with % predicted FEV1. Serum AGE and AGE/sRAGE ratios were high in diabetics but not in CF. A complex, multifaceted approach was used to assess the role of RAGE and its ligands which is fundamental to determining their impact on airway inflammation. There is a clear association between RAGE activity in the airways of CF and CFRD patients that is not evident in the vascular compartment and correlates with lung function, in contrast to diabetes. This strongly suggests a role for RAGE in contributing to the inflammatory overdrive seen in CF and to a greater extent in CFRD.
Product Description
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder