Merck
  • Temporal response properties of koniocellular (blue-on and blue-off) cells in marmoset lateral geniculate nucleus.
Temporal response properties of koniocellular (blue-on and blue-off) cells in marmoset lateral geniculate nucleus.
Journal of neurophysiology (2014-06-13)
A N J Pietersen, S K Cheong, S G Solomon, C Tailby, P R Martin
ABSTRACT
Visual perception requires integrating signals arriving at different times from parallel visual streams. For example, signals carried on the phasic-magnocellular (MC) pathway reach the cerebral cortex pathways some tens of milliseconds before signals traveling on the tonic-parvocellular (PC) pathway. Visual latencies of cells in the koniocellular (KC) pathway have not been specifically studied in simian primates. Here we compared MC and PC cells to "blue-on" (BON) and "blue-off" (BOF) KC cells; these cells carry visual signals originating in short-wavelength-sensitive (S) cones. We made extracellular recordings in the lateral geniculate nucleus (LGN) of anesthetized marmosets. We found that BON visual latencies are 10-20 ms longer than those of PC or MC cells. A small number of recorded BOF cells (n = 7) had latencies 10-20 ms longer than those of BON cells. Within all cell groups, latencies of foveal receptive fields (<10° eccentricity) were longer (by 3-8 ms) than latencies of peripheral receptive fields (>10°). Latencies of yellow-off inputs to BON cells lagged the blue-on inputs by up to 30 ms, but no differences in visual latency were seen on comparing marmosets expressing dichromatic ("red-green color-blind") or trichromatic color vision phenotype. We conclude that S-cone signals leaving the LGN on KC pathways are delayed with respect to signals traveling on PC and MC pathways. Cortical circuits serving color vision must therefore integrate across delays in (red-green) chromatic signals carried by PC cells and (blue-yellow) signals carried by KC cells.
Sigma-Aldrich
Sodium DL-lactate, ReagentPlus®, ≥99.0% (NT)
Sigma-Aldrich
Sodium DL-lactate solution, syrup, BioReagent, 60 % (w/w), synthetic, suitable for cell culture
Sigma-Aldrich
Sodium DL-lactate solution, syrup, 60 % (w/w), synthetic
Sigma-Aldrich
Sodium DL-lactate solution, synthetic, syrup, BioXtra, suitable for mouse embryo cell culture, 60 % (w/w)