Merck
  • The effects of organic solvents on the membrane-induced fibrillation of human islet amyloid polypeptide and on the inhibition of the fibrillation.
The effects of organic solvents on the membrane-induced fibrillation of human islet amyloid polypeptide and on the inhibition of the fibrillation.
Biochimica et biophysica acta (2014-09-15)
Li Wang, Yang Li, Tong Lu, Fei Li
ABSTRACT
The organic solvent dimethylsulphoxide (DMSO) and 1,1,1,3,3,3-hexafluoro-2-isopropanol (HFIP) have been widely used as a pre-treating agent of amyloid peptides and as a vehicle for water-insoluble inhibitors. These solvents are left in many cases as a trace quantity in bulk and membrane environments with treated amyloid peptides or inhibitors. In the present work, we studied the effects of the two organic solvents on the aggregation behaviors of human islet amyloid polypeptide (hIAPP) and the performances of an all-D-amino-acid inhibitor D-NFGAIL in preventing hIAPP fibrillation both in bulk solution and at phospholipid membrane. We showed that the presence of 1% v/v DMSO or HFIP decreases the rate of fibril formation of hIAPP at the lipid membrane rather than accelerates the fibril formation as what happened in bulk solution. We also showed that the presence of 1% v/v DMSO or HFIP impairs the activity of the inhibitor at the lipid membrane surface dramatically, while it affects the efficiency of the inhibitor in bulk solution slightly. We found that the inhibitor inserts into the lipid membrane more deeply or with more proportion in the presence of the organic solvents than it does in the absence of the organic solvents, which may hinder the binding of the inhibitor to hIAPP at the lipid membrane. Our results suggest that the organic solvents should be used with caution in studying membrane-induced fibrillogenesis of amyloid peptides and in testing amyloid inhibitors under membrane environments to avoid incorrect evaluation to the fibrillation process of amyloid peptides and the activity of inhibitors.
Product Description
Supelco
1,1,1,3,3,3-Hexafluoro-2-propanol, for GC derivatization, LiChropur, ≥99.8%
Sigma-Aldrich
1,1,1,3,3,3-Hexafluoro-2-propanol, ≥99%
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Supelco
Glycerol, analytical standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)