Merck
  • Species identification of Mycobacterium abscessus subsp. abscessus and Mycobacterium abscessus subsp. bolletii using rpoB and hsp65, and susceptibility testing to eight antibiotics.
Species identification of Mycobacterium abscessus subsp. abscessus and Mycobacterium abscessus subsp. bolletii using rpoB and hsp65, and susceptibility testing to eight antibiotics.
International journal of infectious diseases : IJID : official publication of the International Society for Infectious Diseases (2014-06-17)
Wenjuan Nie, Hongfei Duan, Hairong Huang, Yu Lu, Dapeng Bi, Naihui Chu
ABSTRACT
To separate Mycobacterium abscessus subsp. bolletii from Mycobacterium abscessus subsp. abscessus using species identification, and to investigate the in vitro activity of amikacin, cefoxitin, imipenem, levofloxacin, moxifloxacin, clarithromycin, azithromycin, and linezolid against Mycobacterium abscessus. Seventy M. abscessus isolates, previously identified by 16S rRNA sequencing, were further identified by comparative sequence analysis of rpoB and hsp65. Drug susceptibility testing was conducted using the microplate Alamar Blue assay in accordance with Clinical and Laboratory Standards Institute (CLSI) guidelines and interpreted using CLSI breakpoints. Of the 70 strains, 45 (64%) were M. abscessus subsp. abscessus and 25 (36%) were M. abscessus subsp. bolletii. The majority of M. abscessus isolates were susceptible to azithromycin, amikacin, linezolid, and imipenem (M. abscessus subsp. abscessus: 93%, 98%, 93%, and 73%, respectively; M. abscessus subsp. bolletii: 96%, 96%, 80%, and 68%, respectively). Approximately half of the M. abscessus isolates were moderately susceptible to cefoxitin and moxifloxacin (M. abscessus subsp. abscessus 53% and 49%; M. abscessus subsp. bolletii 72% and 68%). Nearly all the M. abscessus isolates were resistant to levofloxacin (M. abscessus subsp. abscessus 96%, M. abscessus subsp. bolletii 100%). Inducible clarithromycin resistance was found in M. abscessus. After 14 days of incubation, 83% M. abscessus subsp. abscessus and 36% M. abscessus subsp. bolletii were resistant to clarithromycin. Using rpoB and hsp65, M. abscessus subsp. bolletii could be distinguished from M. abscessus subsp. abscessus. Amikacin and azithromycin showed excellent activity against M. abscessus in vitro. Imipenem, linezolid, cefoxitin, and moxifloxacin also showed good activity. Levofloxacin was inactive against M. abscessus. Although clarithromycin showed excellent activity against M. abscessus on day 3, inducible resistance occurred, and after 14 days clarithromycin showed little activity against M. abscessus subsp. abscessus, but still had good activity against M. abscessus subsp. bolletii.
Sigma-Aldrich
Azithromycin
Sigma-Aldrich
Levofloxacin, 98.0-102.0% anhydrous basis (HPLC)
Supelco
Levofloxacin, analytical standard
USP
Amikacin, United States Pharmacopeia (USP) Reference Standard
USP
Clarithromycin Identity, United States Pharmacopeia (USP) Reference Standard
Amikacin, European Pharmacopoeia (EP) Reference Standard
Supelco
Clarithromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Clarithromycin, ≥95% (HPLC)
Azithromycin for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Clarithromycin, 96.0-102.0% (HPLC)
USP
Levofloxacin, United States Pharmacopeia (USP) Reference Standard
USP
Azithromycin, United States Pharmacopeia (USP) Reference Standard
USP
Clarithromycin, United States Pharmacopeia (USP) Reference Standard
Clarithromycin, European Pharmacopoeia (EP) Reference Standard
Clarithromycin for peak identification, European Pharmacopoeia (EP) Reference Standard
Azithromycin for system suitability, European Pharmacopoeia (EP) Reference Standard
Azithromycin, European Pharmacopoeia (EP) Reference Standard
USP
Azithromycin Identity, United States Pharmacopeia (USP) Reference Standard