Merck
  • Protective effect of quercetin in ecto-enzymes, cholinesterases, and myeloperoxidase activities in the lymphocytes of rats exposed to cadmium.
Protective effect of quercetin in ecto-enzymes, cholinesterases, and myeloperoxidase activities in the lymphocytes of rats exposed to cadmium.
Molecular and cellular biochemistry (2014-07-30)
Fátima Husein Abdalla, Andréia Machado Cardoso, Roberta Schmatz, Jamile Fabbrin Gonçalves, Jucimara Baldissarelli, Caroline Curry Martins, Daniela Zanini, Lizielle Souza de Oliveira, Pauline da Costa, Victor Camera Pimentel, Luciane Belmonte Pereira, Cibele Lima Lhamas, Maria Rosa Chitolina Schetinger, Vera Maria Morsch, Cinthia Melazzo Andrade Mazzanti
ABSTRACT
The ex vivo and in vitro effects of quercetin on NTPDase, adenosine deaminase (ADA), and acetycholinesterase (AChE) activities in lymphocytes, as well as the effects of quercetin on butyrylcholinesterase (BChE) activity in serum and myeloperoxidase (MPO) activity in plasma were determined in rats. For the ex vivo experiment, animals were orally exposed to Cadmium (Cd) for 45 days. Animals were divided into eight groups: saline/ethanol, saline/Querc 5 mg/kg, saline/Querc 25 mg/kg, saline/Querc 50 mg/kg, Cd/ethanol, Cd/Querc 5 mg/kg, Cd/Querc 25 mg/kg, and Cd/Querc 50 mg/kg. The ex vivo data showed an increase in the ATP and ADP hydrolysis and ADA activity in Cd-exposed rats when compared to the control group. The treatment with quercetin 25 and 50 mg/kg prevented this increase in the ATP and ADP hydrolysis, while the treatment with quercetin 5, 25, and 50 mg/kg prevented the increase in the ADA activity. AChE, BChE, and MPO activities ex vivo presented an increase in the Cd-exposed group when compared to the control group, and the treatment with quercetin 5, 25, and 50 mg/kg prevented this increase caused by Cd exposure. The in vitro experiment showed that quercetin 5, 10, 25, or 50 µM decreased the ADA activity proportionally to the increase of the concentrations of quercetin when compared to the control group. Thus, we can suggest that the quercetin is able to modulate NTPDase, ADA, AChE, and MPO activities and contribute to maintain the levels of ATP, adenosine, and acetylcholine normal, respectively, exhibiting potent pro-inflammatory and anti-inflammatory actions.
Sigma-Aldrich
2-Nitrobenzoic acid, 95%, Contains 3- and 4-isomers
Sigma-Aldrich
Ethanolamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
4-Aminoantipyrine, puriss. p.a., reag. Ph. Eur., ≥99%
Sigma-Aldrich
Ethanolamine, ≥99%
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
Ethanolamine, ACS reagent, ≥99.0%
Supelco
4-Aminoantipyrine, for spectrophotometric det. of H2O2 and phenols, ≥98.0%
Supelco
Aucubin, analytical standard
Sigma-Aldrich
Ethanolamine, liquid, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Ethanolamine, ≥98%
Sigma-Aldrich
4-Aminoantipyrine, reagent grade
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Supelco
Ethanolamine, analytical standard
Sigma-Aldrich
Ethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC/NT)
USP
Quercetin, United States Pharmacopeia (USP) Reference Standard
Supelco
4-Aminoantipyrine, analytical standard
Supelco
Quercetin, Pharmaceutical Secondary Standard; Certified Reference Material
Aucubin, primary reference standard
Sigma-Aldrich
Acetylthiocholine iodide, ≥99.0% (AT)
Sigma-Aldrich
Butyrylthiocholine iodide, puriss., ≥99.0% (AT)
Sigma-Aldrich
Butyrylthiocholine iodide, ≥98%
Sigma-Aldrich
Acetylthiocholine iodide, ≥98% (TLC), powder or crystals
Sigma-Aldrich
Cadmium chloride, technical grade
Sigma-Aldrich
Cadmium chloride, 99.99% trace metals basis
Trolamine impurity A, European Pharmacopoeia (EP) Reference Standard