Merck
  • Knockdown of Burton's tyrosine kinase confers potent protection against sepsis-induced acute lung injury.
Knockdown of Burton's tyrosine kinase confers potent protection against sepsis-induced acute lung injury.
Cell biochemistry and biophysics (2014-06-08)
Panyu Zhou, Bing Ma, Shuogui Xu, Shijie Zhang, Hongtai Tang, Shihui Zhu, Shichu Xiao, Daofeng Ben, Zhaofan Xia
ABSTRACT
Sepsis is a common and critical complication in surgical patients that often leads to multiple organ failure syndrome (MOFS), including acute lung injury (ALI) and acute respiratory distress syndrome (ARDS). Despite intensive supportive care and treatment modalities, the mortality of these patients remains high. In this study, we investigated the role of Burton's tyrosine kinase (BTK), a member of the Btk/Tec family of cytoplasmic tyrosine kinases, in the pathogenesis of sepsis, and evaluated the protective effect of in vivo Btk RNA interference in a mouse model of cecal ligation and puncture (CLP)-induced sepsis. After intratracheal injection of Btk siRNA, the mice were then subjected to CLP to induce sepsis. The results demonstrated that this approach conferred potent protection against sepsis-induced ALI, as evidenced by a significant reduction in pathological scores, epithelial cell apoptosis, pulmonary edema, vascular permeability, and the expression of inflammatory cytokines and neutrophil infiltration in the lung tissues of septic mice. In addition, RNA interference of Btk significantly suppressed p-38 and iNOS signaling pathways in transduced alveolar macrophages in vitro. These results identify a novel role for BTK in lethal sepsis and provide a potential new therapeutic approach to sepsis and ALI.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Mouse IL-6 R ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Rat IL-6 ELISA Kit, for serum, plasma and cell culture supernatants
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Btk
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Rat Interleukin-6 ELISA Kit, for for cell lysate and tissue lysate
Sigma-Aldrich
MISSION® esiRNA, targeting human BTK
Sigma-Aldrich
Mouse IL-6 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Mouse IL-6 ELISA Kit, for cell and tissue lysates