Merck
  • PSP94 contributes to chemoresistance and its peptide derivative PCK3145 represses tumor growth in ovarian cancer.
PSP94 contributes to chemoresistance and its peptide derivative PCK3145 represses tumor growth in ovarian cancer.
Oncogene (2013-11-05)
B-X Yan, J-X Ma, J Zhang, Y Guo, H Riedel, M D Mueller, S C Remick, J J Yu
ABSTRACT
Tumor drug resistance remains a major challenge in the treatment of cancer. Here, we show that Prostatic secretory protein 94 (PSP94) levels are reduced in ovarian cancer patients with high levels of excision repair cross-complementing 1 (ERCC1), a marker for chemoresistance. We find that PSP94 is decreased in an ovarian cancer drug-resistant cell line, and plays an important role in the development of drug resistance in vitro. Our studies indicate that PSP94 can partially reverse drug resistance in mouse tumor models in vivo and that a PSP94 peptide derivative PCK3145 suppresses chemoresistant cancer cell and tumor growth in vitro and in vivo. Our investigation of the involved molecular mechanisms suggests that PSP94 may confer drug resistance by modulating the Lin28b/Let-7 signaling pathway. We introduce PSP94 and its peptide derivative PCK3145 as potential target to reverse chemoresistance in ovarian cancer and have begun to identify their relevant molecular targets in specific signaling pathways.
Product Description
Supelco
Ethanol, standard for GC
Pricing and availability is not currently available.
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Pricing and availability is not currently available.
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethanol
Pricing and availability is not currently available.
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human LIN28B
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Pricing and availability is not currently available.
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Lin28b
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Pricing and availability is not currently available.
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.