Merck
  • Anti-obesogenic effects of calcium prevent changes in the GLP-1 profile in adult rats primed by early weaning.
Anti-obesogenic effects of calcium prevent changes in the GLP-1 profile in adult rats primed by early weaning.
Molecular nutrition & food research (2015-01-13)
Fernanda Torres Quitete, Jessica Lopes Nobre, Nayara Peixoto-Silva, Egberto Gaspar de Moura, Patricia Cristina Lisboa, Elaine de Oliveira
ABSTRACT
Gut peptides regulate appetite and adipogenesis. Early weaning (EW) leads to later development of obesity that can be prevented by calcium supplementation. We evaluated gut peptides that may have a role in the establishment of this dysfunction. At birth, lactating Wistar rats were separated in: EW, lactating rats involved with a bandage interrupting the lactation during the last 4 days of standard lactation, and C (control) dams whose pups had free access to milk during throughout lactation. At 120 days old, half of EW group received calcium supplementation (EWCa); EW and C received standard diet. At 21 days old, EW presented higher glucagon-like peptide 1 (GLP-1) in plasma and glucagon-like peptide 1 receptor (GLP1-R) in adipose tissue and hypothalamus, but lower GLP-1 and GLP1-R in the gut. At 180 days old, GLP-1 response to food intake was blunted in EW and restored by calcium. GLP-1 in the gut was lower in EW and its receptor was lower in adipose tissue, and GLP1-R was higher in the gut of calcium EW group. Thus, EW had short- and long-term effects upon GLP-1 profile, which may have contributed to obesity development, hyperphagia, and insulin resistance due to its adipogenic and appetite control roles. Calcium supplementation was able to prevent most of the changes in GLP-1 caused by EW.
Product Description
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
USP
Calcium Carbonate (AS), United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
o-Xylene, analytical standard
Sigma-Aldrich
o-Xylene, anhydrous, 97%