Merck
  • Unsaturated free fatty acids: a potential biomarker panel for early detection of gastric cancer.
Unsaturated free fatty acids: a potential biomarker panel for early detection of gastric cancer.
Biomarkers : biochemical indicators of exposure, response, and susceptibility to chemicals (2014-10-31)
Yaping Zhang, Ling Qiu, Yanmin Wang, Chengyan He, Xuzhen Qin, Yujie Liu, Zhili Li
ABSTRACT
Changes in the levels of free fatty acids (FFAs) are closely associated with physiological status. Serum levels of C16:1, C18:3, C18:2, C18:1, C20:4, and C22:6 in 164 gastric cancer (GC) patients and 111 benign gastric disease (BGD) patients were significantly decreased compared with 252 healthy controls. Receiver operating characteristic analysis showed that the biomarker panel including C16:1, C18:3, C18:2, C20:4, and C22:6 presents a high diagnostic ability to differentiate early-stage GC patients from healthy controls plus BGD patients, with a sensitivity of 80.6% and a specificity of 72.7%.
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Heneicosanoic acid, analytical standard
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology