Merck
  • Simultaneous determination of perfluorinated compounds and their potential precursors in mussel tissue and fish muscle tissue and liver samples by liquid chromatography-electrospray-tandem mass spectrometry.
Simultaneous determination of perfluorinated compounds and their potential precursors in mussel tissue and fish muscle tissue and liver samples by liquid chromatography-electrospray-tandem mass spectrometry.
Journal of chromatography. A (2015-02-25)
I Zabaleta, E Bizkarguenaga, A Prieto, M Ortiz-Zarragoitia, L A Fernández, O Zuloaga
ABSTRACT
An analytical method for the simultaneous determination in fish liver and muscle tissue and mussel samples of 14 perfluorinated compounds (PFCs), including three perfluoroalkylsulfonates (PFSAs), seven perfluorocarboxylic acids (PFCAs), three perfluorophosphonic acids (PFPAs) and perfluorooctanesulfonamide (PFOSA), and 10 potential precursors, including four polyfluoroalkyl phosphates (PAPs), four fluorotelomer saturated acids (FTCAs) and two fluorotelomer unsaturated acids (FTUCAs), was developed in the present work. Different clean-up strategies by means of solid-phase extraction (SPE) using a mix-mode weak anion exchanger (WAX), reverse phase Envi-Carb or a combination of them was optimized and evaluated for the clean-up of focused ultrasonic solid-liquid (FUSLE) extracts before the analysis by liquid chromatography-triple quadrupole tandem mass spectrometry (LC-MS/MS). Mix-mode WAX coupled in-line to Envi-Carb was finally selected since it rendered the cleanest extracts and minimum matrix effect. The FUSLE-SPE-LC-MS/MS methodology was validated in terms of recovery, precision and method detection limits (MDLs). Apparent recovery values in the 65-116%, 59-119% and 67-126% range and MDLs in the 0.1-2.7 ng/g, 0.1-3.8 ng/g and 0.2-3.1ng/g range were obtained for liver, mussel and fish muscle tissue samples, respectively. The method developed was applied to the analysis of grey mullet liver (Chelon labrosus) and mussel (Mytilus galloprovincialis) samples from the Basque Coast (North of Spain) and Yellowfin tuna muscle tissue (Thunnus albacares) samples from the Indian Ocean. To the best of our knowledge this is the first method that describes the simultaneous determination of 14 PFCs and 10 potential precursors in fish liver, fish muscle tissue and mussel samples. Besides, this is the first time that 8:2 monosubstituted polyfluorodecyl phosphate (8:2 monoPAP) and 8:2 disubstituted polyfluorodecyl phosphate (8:2 diPAP) were detected in mussel and tuna samples, respectively.
Sigma-Aldrich
Carbon, mesoporous, nanopowder, less than 500 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Carbon, mesoporous
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Supelco
tert-Butyl Methyl Ether, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, steam activated, rod
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium acetate solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, BioUltra, ≥99.0%
Sigma-Aldrich
Methanol, anhydrous, 99.8%