Merck
  • Study of the matrix effects and sample dilution influence on the LC-ESI-MS/MS analysis using four derivatization reagents.
Study of the matrix effects and sample dilution influence on the LC-ESI-MS/MS analysis using four derivatization reagents.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-08-12)
Maarja-Liisa Oldekop, Koit Herodes, Riin Rebane
ABSTRACT
For liquid chromatographic analysis of amino acids involving derivatization and mass-spectrometric detection, it becomes more important to evaluate the presence of matrix effects in complex samples. This is somewhat complicated for amino acid analysis where analyte free sample matrix is often unavailable. In this work, matrix effects were investigated using post-column infusion method for 9-fluorenylmethyl chloroformate (FMOC-Cl) derivatives of β-Ala, Gly and Phe and diethyl ethoxymethylenemalonate (DEEMM) derivative of β-Ala. While for DEEMM derivatives, the main signal suppression was due to the borate buffer, in case of FMOC-Cl, other FMOC-derivatives caused signal suppression. Analysis of amino acids in tea and honey with DEEMM, FMOC-Cl, p-N,N,N-trimethylammonioanilyl N'-hydroxysuccinimidyl carbamate iodide (TAHS) and dansyl chloride (DNS) showed that amino acid concentrations found with different reagents do not agree well. Sample dilution experiments indicated that the sample matrix affected the analysis results obtained with DEEMM the least, but with FMOC-Cl, TAHS and DNS, sample dilution had an influence on the results. When sample dilution and extrapolative dilution approach were applied on the latter results, an agreement of amino acid concentrations measured with different reagents was achieved within relative standard deviation (RSD) of 22% for most cases.
Product Description
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Supelco
L-Proline, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Histidine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Histidine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Proline, Pharmaceutical Secondary Standard; Certified Reference Material
USP
L-Proline, United States Pharmacopeia (USP) Reference Standard
SAFC
L-Aspartic acid
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)