Merck
  • Free fatty acid receptor 2, a candidate target for type 1 diabetes, induces cell apoptosis through ERK signaling.
Free fatty acid receptor 2, a candidate target for type 1 diabetes, induces cell apoptosis through ERK signaling.
Journal of molecular endocrinology (2014-10-10)
Guojun Shi, Chen Sun, Weiqiong Gu, Minglan Yang, Xiaofang Zhang, Nan Zhai, Yan Lu, Zhijian Zhang, Peishun Shou, Zhiguo Zhang, Guang Ning
ABSTRACT
Recent reports have highlighted the roles of free fatty acid receptor 2 (FFAR2) in the regulation of metabolic and inflammatory processes. However, the potential function of FFAR2 in type 1 diabetes (T1D) remains unexplored. Our results indicated that the mRNA level of FFAR2 was upregulated in peripheral blood mononuclear cells of T1D patients. The human FFAR2 promoter regions were cloned, and luciferase reporter assays revealed that NFκB activation induced FFAR2 expression. Furthermore, we showed that FFAR2 activation by overexpression induced cell apoptosis through ERK signaling. Finally, treatment with the FFAR2 agonists acetate or phenylacetamide 1 attenuated the inflammatory response in multiple-low-dose streptozocin-induced diabetic mice, and improved the impaired glucose tolerance. These results indicate that FFAR2 may play a protective role by inducing apoptosis of infiltrated macrophage in the pancreas through its feedback upregulation and activation, thus, in turn, improving glucose homeostasis in diabetic mice. These findings highlight FFAR2 as a potential therapeutic target of T1D, representing a link between immune response and glucose homeostasis.
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
Methylglyoxal solution, ~40% in H2O
Sigma-Aldrich
Fluorescein diacetate, used as cell viability stain
Sigma-Aldrich
Methylglyoxal solution, technical, ~40% in H2O
Sigma-Aldrich
Propidium iodide solution
USP
Sodium acetate, United States Pharmacopeia (USP) Reference Standard
Supelco
Sodium propionate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium propionate, ≥99.0%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Pyruvaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium propionate, FCC
Diacetylfluorescein, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium propionate, meets analytical specification of NF, 99-100.5% (ex dried subst.)
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium propionate, ≥99.0%, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%