Merck
  • Preparation of risedronate nanoparticles by solvent evaporation technique.
Preparation of risedronate nanoparticles by solvent evaporation technique.
Molecules (Basel, Switzerland) (2014-11-07)
Eliska Vaculikova, Daniela Placha, Martin Pisarcik, Pavlina Peikertova, Katerina Dedkova, Ferdinand Devinsky, Josef Jampilek
ABSTRACT
One approach for the enhancement of oral drug bioavailability is the technique of nanoparticle preparation. Risedronate sodium (Biopharmaceutical Classification System Class III) was chosen as a model compound with high water solubility and low intestinal permeability. Eighteen samples of risedronate sodium were prepared by the solvent evaporation technique with sodium dodecyl sulfate, polysorbate, macrogol, sodium carboxymethyl cellulose and sodium carboxymethyl dextran as nanoparticle stabilizers applied in three concentrations. The prepared samples were characterized by dynamic light scattering and scanning electron microscopy. Fourier transform mid-infrared spectroscopy was used for verification of the composition of the samples. The particle size of sixteen samples was less than 200 nm. Polysorbate, sodium carboxymethyl dextran and macrogol were determined as the most favourable excipients; the particle size of the samples of risedronate with these excipients ranged from 2.8 to 10.5 nm.
Sigma-Aldrich
Risedronate sodium, ≥97% (HPLC)
Sigma-Aldrich
Argon, ≥99.998%
Sigma-Aldrich
Argon-40Ar, 99.95 atom %
USP
Risedronate sodium, United States Pharmacopeia (USP) Reference Standard