Merck
  • Behaviour and ultrastructure of human bone marrow-derived mesenchymal stem cells immobilised in alginate-poly-l-lysine-alginate microcapsules.
Behaviour and ultrastructure of human bone marrow-derived mesenchymal stem cells immobilised in alginate-poly-l-lysine-alginate microcapsules.
Journal of microencapsulation (2014-04-29)
Noha Attia, Edorta Santos, Hala Abdelmouty, Samia Arafa, Nahed Zohdy, Rosa María Hernández, Gorka Orive, José Luis Pedraz
ABSTRACT
Human bone marrow mesenchymal stem cells (hBM-MSCs) show a great promise for the treatment of a variety of diseases. Despite the previous trials to encapsulate hBM-MSCs in alginate-poly-l-lysine-alginate (APA) systems, the various changes that follow immobilisation have not been ascertained yet. Determine the various consequences derived from entrapment on cell behaviour, putting special emphasis on the ultrastructure. hBM-MSCs were immobilised in APA microcapsules to further characterise their viability, metabolic activity, proliferation, VEGF-secretability, and morphology. The VEGF produced by monolayer hBM-MSCs increased significantly 1 d post-encapsulation, and was maintained for at least 4  weeks. TEM imaging of cells revealed well preserved ultrastructure indicating protein synthesis and high metabolic activity. Although APA microencapsulation did not support 100% of fully viable hBM-MSCs for long-term cultures, it was conceived to enhance both VEGF secretion and metabolic activity while not losing their stemness characteristics.
Product Description
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Indomethacin, European Pharmacopoeia (EP) Reference Standard
Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Diethyl azodicarboxylate solution, purum, ~40% in toluene (H-NMR)
USP
Indomethacin, United States Pharmacopeia (USP) Reference Standard
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Dexamethasone, British Pharmacopoeia (BP) Assay Standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrogen bromide solution, 33 wt. % in acetic acid
Sigma-Aldrich
Hydrobromic acid, reagent grade, 48%
Sigma-Aldrich
Hydrobromic acid, ACS reagent, 48%
Sigma-Aldrich
Hydrobromic acid, 48 wt. % in H2O, ≥99.99%