Merck
  • Key changes in wine aroma active compounds during bottle storage of Spanish red wines under different oxygen levels.
Key changes in wine aroma active compounds during bottle storage of Spanish red wines under different oxygen levels.
Journal of agricultural and food chemistry (2014-10-07)
Vicente Ferreira, Mónica Bueno, Ernesto Franco-Luesma, Laura Culleré, Purificación Fernández-Zurbano
ABSTRACT
Samples from 16 Spanish red wines have been stored for 6 months at 25 °C under different levels of oxygen (0-56 mg/L). Amino acids, metals, and phenolic compounds were analyzed and related to the production or depletion of key oxidation- and reduction-related aroma compounds. Oxidation brings about sensory-relevant increases in Strecker aldehydes, 1-octen-3-one, and vanillin. Formation of Strecker aldehydes correlates to the wine content on the corresponding amino acid precursor, Zn, and caffeic acid ethyl ester and negatively to some flavonols and anthocyanin derivatives. Formation of most carbonyls correlates to wine-combined SO2, suggesting that part of the increments are the result of the release of aldehydes forming bisulfite combinations once SO2 is oxidized. Methanethiol (MeSH) and dimethylsulfide (DMS), but not H2S levels, increase during storage. MeSH increments correlate to methionine levels and proanthocyanidins and negatively to resveratrol and aluminum. H2S, MeSH, and DMS levels all decreased with oxidation, and for the latter two, there are important effects of Mn and pH, respectively.
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Supelco
Diethyl ether, analytical standard
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
Diethyl ether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Diethyl ether, suitable for HPLC, ≥99.9%, inhibitor-free
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Diethyl ether, reagent grade, ≥98%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Pentane, analytical standard
Supelco
Hexane, analytical standard
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
Pentane, anhydrous, ≥99%
Supelco
Dichloromethane, analytical standard
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer