Merck
  • Determination of total iodine in serum and urine samples by ion chromatography with pulsed amperometric detection - studies on analyte loss, optimization of sample preparation procedures, and validation of analytical method.
Determination of total iodine in serum and urine samples by ion chromatography with pulsed amperometric detection - studies on analyte loss, optimization of sample preparation procedures, and validation of analytical method.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-06-10)
Anna Błażewicz, Maria Klatka, Wojciech Dolliver, Ryszard Kocjan
ABSTRACT
A fast, accurate and precise ion chromatography method with pulsed amperometric detection was applied to evaluate a variety of parameters affecting the determination of total iodine in serum and urine of 81 subjects, including 56 obese and 25 healthy Polish children. The sample pretreatment methods were carried out in a closed system and with the assistance of microwaves. Both alkaline and acidic digestion procedures were developed and optimized to find the simplest combination of reagents and the appropriate parameters for digestion that would allow for the fastest, least time consuming and most cost-effective way of analysis. A good correlation between the certified and the measured concentrations was achieved. The best recoveries (96.8% for urine and 98.8% for serum samples) were achieved using 1ml of 25% tetramethylammonium hydroxide solution within 6min for 0.1ml of serum/urine samples. Using 0.5ml of 65% nitric acid solution the best recovery (95.3%) was obtained when 7min of effective digestion time was used. Freeze-thaw stability and long-term stability were checked. After 24 weeks 14.7% loss of iodine in urine, and 10.9% in serum samples occurred. For urine samples, better correlation (R(2)=0.9891) of various sample preparation procedures (alkaline digestion and application of OnGuard RP cartidges) was obtained. Significantly lower iodide content was found in samples taken from obese children. Serum iodine content in obese children was markedly variable in comparison with the healthy group, whereas the difference was less evident when urine samples were analyzed. The mean content in serum was 59.12±8.86μg/L, and in urine 98.26±25.93 for obese children when samples were prepared by the use of optimized alkaline digestion reinforced by microwaves. In healthy children the mean content in serum was 82.58±6.01μg/L, and in urine 145.76±31.44μg/L.
Sigma-Aldrich
Potassium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Tetramethylammonium hydroxide solution, ACS reagent
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in methanol
Sigma-Aldrich
Potassium iodide, tested according to Ph. Eur.
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in H2O
Sigma-Aldrich
Potassium iodide, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Potassium iodide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 10 wt. % in H2O
Sigma-Aldrich
Potassium iodide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium iodide, suitable for plant cell culture
Sigma-Aldrich
Ammonia, puriss., anhydrous, ≥99.95%
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Potassium iodide, puriss., meets analytical specification of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance)
Supelco
Potassium Iodide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium iodide, ReagentPlus®, 99%
Sigma-Aldrich
Potassium iodide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium iodide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Ammonia solution, 2.0 M in ethanol
Sigma-Aldrich
Ammonia solution, 7 N in methanol
Sigma-Aldrich
Ammonia solution, 2.0 M in isopropanol
Sigma-Aldrich
Potassium iodide, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonia solution, 0.4 M in dioxane
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Supelco
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Nitric acid, ACS reagent, 70%