Merck
  • Overexpression of Lhx8 inhibits cell proliferation and induces cell cycle arrest in PC12 cell line.
Overexpression of Lhx8 inhibits cell proliferation and induces cell cycle arrest in PC12 cell line.
In vitro cellular & developmental biology. Animal (2014-12-06)
Haoming Li, Jianbing Qin, Guohua Jin, Linqing Zou, Jinhong Shi, Xiao Han, Xiang Cheng, Xinhua Zhang
ABSTRACT
LIM-homeobox genes play a pivotal function in tissue patterning and differentiation, Lhx8 is a member of LIM-homeobox gene family, and it is selectively expressed in embryonic basal forebrain and is a key factor for the determination of cholinergic cells fate. However, besides cholinergic differentiation, little is known about the potential role of Lhx8 in cell biology. In this study, we transfected Lhx8 complementary DNA (cDNA) into PC12 cell line using lentiviral vectors to acquire the cells which stably expressed high level of Lhx8, and we provide the experimental evidence that overexpression of Lhx8 inhibits cell proliferation and induces cell cycle arrest but not apoptosis in vitro. In conclusion, besides cholinergic differentiation, our results suggest that Lhx8 also plays as a suppressor gene of proliferation in cell biology.
Product Description
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Ethanol, for residue analysis
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology