Merck
  • Technical brief: Optimized pipeline for isolation of high-quality RNA from corneal cell subpopulations.
Technical brief: Optimized pipeline for isolation of high-quality RNA from corneal cell subpopulations.
Molecular vision (2014-06-19)
Chris Bath, Trine Fink, Henrik Vorum, Jesper Hjortdal, Vladimir Zachar
ABSTRACT
Attempts to determine the transcriptional profile of discrete subsets of limbal epithelial cells in situ using laser capture microdissection (LCM) face two major challenges. First, the transcriptional profile of cells within a tissue may rapidly change as the tissue is excised and exposed to cold ischemia. Second, there is a risk of degradation of the RNA as the cellular compartment is separated from the remaining tissue. An optimized protocol for LCM of corneal epithelium is presented to address these issues. Experiments using porcine eye globes were carried out to determine both optimal procedures and settings for tissue harvest, transport, storage, histology, LCM, and RNA isolation. The optimized protocol was validated using human corneal epithelium. To facilitate preservation of the gene expression profile, we have developed a mechanical tool for dissection of cornea that, in combination with flash freezing, enables tissue to be stored within 5 min of enucleation of the eye. Furthermore, we describe how RNA from limbal crypt cells may be obtained using a procedure involving cryosectioning, histological staining, and LCM. In this paper, we describe an optimized method for isolating high-quality RNA from cellular subpopulations confined to the limbal crypts of the cornea. The procedure yields RNA in amounts and quality suitable for downstream gene expression analyses, such as microarrays or next generation sequencing.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Xylazine, ≥99%
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol, for residue analysis
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology