Merck
  • Microbiological, physico-chemical and proteolytic changes in a Spanish blue cheese during ripening (Valdeón cheese).
Microbiological, physico-chemical and proteolytic changes in a Spanish blue cheese during ripening (Valdeón cheese).
Food chemistry (2014-08-31)
I Diezhandino, D Fernández, L González, P L H McSweeney, J M Fresno
ABSTRACT
The aim of this work was to study the microbiological, physico-chemical and proteolytic changes in Valdeón blue-veined cheese during ripening. Eight replicas of cheese were produced and a total of 48 cheeses were analysed. Lactic acid bacteria, mainly lactococci, were the predominant flora during the early stages of ripening, gradually being replaced by moulds and yeasts (8 log units). Enterococci and Enterobacteriaceae counts were very low or zero. This variety was characterised by a total solids content of 61.80g per 100g(-1) of cheese, a salt/moisture ratio of 8.92g salt per 100g(-1) moisture, a pH of 6.4-7.6 and a water activity of 0.917. At the end of ripening, primary and secondary proteolysis were very high, which resulted in an almost total degradation of αs1- and β-casein (approximately 90%). The peptide profile of the aqueous soluble extracts at pH 4.6 showed great complexity during ripening.
Product Description
Millipore
Oxytetra Selective Supplement, suitable for microbiology
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Mannitol, European Pharmacopoeia (EP) Reference Standard
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
Esculin hydrate, ≥98%
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
Esculin hydrate, ≥98.0% (HPLC)
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Supelco
Oxytetracycline, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Oxytetracycline, United States Pharmacopeia (USP) Reference Standard