Merck
  • Multi-residue method for determination of 238 pesticides in Chinese cabbage and cucumber by liquid chromatography-tandem mass spectrometry: comparison of different purification procedures.
Multi-residue method for determination of 238 pesticides in Chinese cabbage and cucumber by liquid chromatography-tandem mass spectrometry: comparison of different purification procedures.
Journal of agricultural and food chemistry (2014-11-08)
Mei-Ai Zhao, Ya-Nan Feng, Yong-Zhe Zhu, Jeong-Han Kim
ABSTRACT
This paper describes the comparison of five sample cleanup procedures for the determination of 238 pesticides via triple quadrupole liquid chromatography-tandem mass spectrometry (LC-MS/MS, with only 10 min of chromatographic running time) in Chinese cabbage and cucumber. Samples were extracted with a quick, easy, cheap, effective, rugged, and safe (QuECHERS) preparation method and cleanup with different sorbents, including primary secondary amine (PSA), multi-walled carbon nanotubes (MWCNTs), and polystyrene (PLS), to find out the most suitable cleanup methods for Chinese cabbage and cucumber. The recovery and matrix effect were evaluated by monitoring the main parameters in one group of 238 pesticides at the spiked level of 8 and 40 μg/kg. In Chinese cabbage, when PSA dispersive solid-phase extraction (D-SPE) was applied, recoveries of 183 pesticides ranged between 70 and 120% with relative standard deviation (RSD) values lower than 20% at a spiked level of 40 μg/kg, indicating the effectiveness of the purification step. In cucumber, 203 pesticides were in the 70-120% recovery range with good reproducibility by PSA mini-cartridge column cleanup at a spiked level of 40 μg/kg and RSD values were generally below 20%. The limits of quantitation [LOQs; signal-to-noise (S/N) = 10] were in the range of 0.16-10.20 μg/kg for Chinese cabbage and 0.06-21.06 μg/kg for cucumber, while the limits of detection (LODs; S/N = 3) were between 0.05 and 3.06 μg/kg and between 0.02 and 6.32 μg/kg in Chinese cabbage and cucumber, respectively. The proposed methods that might be applied for the multi-residue analysis in Chinese cabbage and cucumber are contributed to their rapid speed and good recoveries.
Product Description
Supelco
Benthiavalicarb isopropyl, PESTANAL®, analytical standard
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Supelco
Trichlorfon, PESTANAL®, analytical standard
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Methanol solution, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Magnesium sulfate solution, 1.00 M±0.04 M
Sigma-Aldrich
Magnesium sulfate solution, BioUltra
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Supelco
Triflumuron, PESTANAL®, analytical standard
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Supelco
Terbuthylazine, PESTANAL®, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
SAFC
Sodium chloride solution, 5 M