Merck
  • Use of graphite as a highly reversible electrode with superior cycle life for sodium-ion batteries by making use of co-intercalation phenomena.
Use of graphite as a highly reversible electrode with superior cycle life for sodium-ion batteries by making use of co-intercalation phenomena.
Angewandte Chemie (International ed. in English) (2014-07-25)
Birte Jache, Philipp Adelhelm
ABSTRACT
Although being the standard anode material in lithium-ion batteries (LIBs), graphite so far is considered to fail application in sodium-ion batteries (NIBs) because the Na-C system lacks suitable binary intercalation compounds. Here we show that this limitation can be circumvented by using co-intercalation phenomena in a diglyme-based electrolyte. The resulting compound is a stage-I ternary intercalation compound with an estimated stoichiometry of Na(diglyme)2C20. Highlights of the electrode reaction are its high energy efficiency, the small irreversible loss during the first cycle, and a superior cycle life with capacities close to 100 mAh g(-1) for 1000 cycles and coulomb efficiencies >99.87%. A one-to-one comparison with the analogue lithium-based cell shows that the sodium-based system performs better and also withstands higher currents.
Sigma-Aldrich
Lithium-7Li, ≥99.8 atom % 7Li, ≥99.8% (CP)
Sigma-Aldrich
Sodium triflate solution, in Bis(2-methoxyethyl) ether, 1.0 M NaTfO in diglyme, battery grade
Sigma-Aldrich
Lithium, rod, diam. 12.7 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.75 mm × 45 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.38 mm × 23 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, granular, 4-10 mesh particle size, high sodium, 99% (metals basis)
Sigma-Aldrich
Lithium, wire, diam. 3.2 mm, in mineral oil, ≥98%
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.75 mm × 19 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, wire (in mineral oil), diam. 3.2 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, granular, 99% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 1.5 mm × 100 mm, 99.9% trace metals basis
Lithium, foil, 25x100mm, thickness 0.6mm, as rolled, 99.9%
Sigma-Aldrich
Lithium, foil, not light tested, 38x500mm, thickness 0.20mm, as rolled, 99.9%
Lithium, foil, not light tested, 38x200mm, thickness 0.20mm, as rolled, 99.9%