Merck
  • Development of a LC-MS/MS method for the simultaneous screening of seven water-soluble vitamins in processing semi-coarse wheat flour products.
Development of a LC-MS/MS method for the simultaneous screening of seven water-soluble vitamins in processing semi-coarse wheat flour products.
Analytical and bioanalytical chemistry (2015-02-24)
Eric Nurit, Bernard Lyan, Agnès Piquet, Gérard Branlard, Estelle Pujos-Guillot
ABSTRACT
Wheat is the second largest crop cultivated around the world and constitutes a major part of the daily diet in Europe. It is therefore important to determine the content of micronutrient in wheat and wheat-based food products to define the contribution of wheat-based foods to the nutrition of the consumers. The aim of the present work was to develop a simple and rapid method based on liquid chromatography tandem mass spectrometry (LC-MS/MS) for the simultaneous determination of seven water-soluble vitamins in various wheat-based food materials. The vitamins present in the test material were separated in less than 15 min by using a reverse-phase C18 column, and analyzed by positive ion electrospray selected reaction monitoring MS/MS. The MS response for all the vitamins was linear over the working range (0.05 to 9 μg/mL) with correlation coefficients ranging between 0.991 and 1. Limits of quantification in the different food materials ranged from 0.09 to 3.5 μg/g. Intra-day and inter-day precision were found satisfactory. The developed method was applied for the simultaneous analysis of the water-soluble vitamin natural content of different semi-coarse wheat flours and in their corresponding baking products.
Product Description
Supelco
Riboflavin (B2), analytical standard
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethanol, for residue analysis
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, for molecular biology
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%