Merck
  • Triple subconjunctival bevacizumab injection for early corneal recurrent pterygium: one-year follow-up.
Triple subconjunctival bevacizumab injection for early corneal recurrent pterygium: one-year follow-up.
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics (2014-11-05)
Angel Nava-Castañeda, Isabel Ulloa-Orozco, Lilia Garnica-Hayashi, Joaquín Hernandez-Orgaz, Maria Carmen Jimenez-Martinez, Yonathan Garfias
ABSTRACT
The aim of the study was to evaluate the effect of 3 subconjunctival bevacizumab injections in patients with an early corneal pterygium recurrence. This study was a nonrandomized single center trial. Patients with an early corneal pterygium recurrence were selected. All patients received 3 subconjunctival bevacizumab (2.5 mg/0.1 mL) injections (basal, 2 and 4 weeks) in the recurrence area of the pterygium. The corneal and corneal-conjunctival neovascularization areas and the corneal opacification area of each pterygium were determined using digital slit lamp pictures. Thirty-eight patients were enrolled into the study; all patients were injected within 3 months of the diagnosed pterygium recurrence. Interestingly, the bevacizumab injections had a significant effect (P<0.05) on the reduction of corneal, corneal-conjunctival area of neovascularization determined as pixels and on the corneal opacification area determined as mm(2) when comparing the basal values, to the values obtained after 15 days, 1 month, 3 months, 6 months, and 12 months after injections. The vascularized area in all recurrent pterygia and the corneal opacification area with this triple regimen of subconjunctival bevacizumab injections were reduced, which remained until the end of the study. These results suggest that bevacizumab subconjunctival injections could be useful to treat recurrent pterygium.
Sigma-Aldrich
Tobramycin, Aminoglycoside antibiotic
Sigma-Aldrich
Tetracaine hydrochloride, ≥99%
Tetracaine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Tetracaine for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Tobramycin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Tetracaine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Tetracaine hydrochloride, United States Pharmacopeia (USP) Reference Standard
USP
Tobramycin, United States Pharmacopeia (USP) Reference Standard
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Tobramycin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Dexamethasone, British Pharmacopoeia (BP) Assay Standard