Merck
  • 'Stealth' lipid-based formulations: poly(ethylene glycol)-mediated digestion inhibition improves oral bioavailability of a model poorly water soluble drug.
'Stealth' lipid-based formulations: poly(ethylene glycol)-mediated digestion inhibition improves oral bioavailability of a model poorly water soluble drug.
Journal of controlled release : official journal of the Controlled Release Society (2014-07-25)
Orlagh M Feeney, Hywel D Williams, Colin W Pouton, Christopher J H Porter
ABSTRACT
For over 20years, stealth drug delivery has been synonymous with nanoparticulate formulations and intravenous dosing. The putative determinants of stealth in these applications are the molecular weight and packing density of a hydrophilic polymer (commonly poly(ethylene glycol) (PEG)) that forms a steric barrier at the surface of the nanoparticle. The current study examined the potential translation of the concepts learned from stealth technology after intravenous administration to oral drug delivery and specifically, to enhance drug exposure after administration of oral lipid-based formulations (LBFs) containing medium-chain triglycerides (MCT). MCT LBFs are rapidly digested in the gastrointestinal tract, typically resulting in losses in solubilisation capacity, supersaturation and drug precipitation. Here, non-ionic surfactants containing stealth PEG headgroups were incorporated into MCT LBFs in an attempt to attenuate digestion, reduce precipitation risk and enhance drug exposure. Stealth capabilities were assessed by measuring the degree of digestion inhibition that resulted from steric hindrance of enzyme access to the oil-water interface. Drug-loaded LBFs were assessed for maintenance of solubilising capacity during in vitro digestion and evaluated in vivo in rats. The data suggest that the structural determinants of stealth LBFs mirror those of parenteral formulations, i.e., the key factors are the molecular weight of the PEG in the surfactant headgroup and the packing density of the PEG chains at the interface. Interestingly, the data also show that the presence of labile ester bonds within a PEGylated surfactant also impact on the stealth properties of LBFs, with digestible surfactants requiring a PEG Mw of ~1800g/mol and non-digestible ether-based surfactants ~800g/mol to shield the lipidic cargo. In vitro evaluation of drug solubilisation during digestion showed stealth LBFs maintained drug solubilisation at or above 80% of drug load and reduced supersaturation in comparison to digestible counterparts. This trend was also reflected in vivo, where the relative bioavailability of drug after administration in two stealth LBFs increased to 120% and 182% in comparison to analogous digestible (non-stealth) formulations. The results of the current study indicate that self-assembled "stealth" LBFs have potential as a novel means of improving LBF performance.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Ethylene sulfite, 98%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, ≥99%
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Progesterone for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethylene sulfite, ≥99.0%
Progesterone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Progesterone, European Pharmacopoeia (EP) Reference Standard
USP
Progesterone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Supelco
Progesterone, VETRANAL®, analytical standard
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets