Merck
  • A novel segmental challenge model for bleomycin-induced pulmonary fibrosis in sheep.
A novel segmental challenge model for bleomycin-induced pulmonary fibrosis in sheep.
Experimental lung research (2014-12-23)
Louise Organ, Barbara Bacci, Emmanuel Koumoundouros, Garry Barcham, Wayne Kimpton, Cameron J Nowell, Chrishan Samuel, Ken Snibson
ABSTRACT
Idiopathic Pulmonary fibrosis (IPF) is a fatal respiratory disease, characterized by a progressive fibrosis and worsening lung function. While the outcomes of recent clinical trials have resulted in therapies to slow the progression of the disease, there is still a need to develop alternative therapies, which are able to prevent fibrosis. This study uses a segmental lung infusion of bleomycin (BLM) to investigate pulmonary fibrosis in a physiologically relevant large animal species. Two separate lung segments in eight sheep received two fortnightly challenges of either 3U or 30U BLM per segment, and a third segment received saline (control). Lung function was assessed using a wedged-bronchoscope procedure. Bronchoalveolar lavage fluid and lung tissue were assessed for inflammation, fibrosis and collagen content two weeks after the final dose of BLM. Instillation of both BLM doses resulted in prominent fibrosis in the treated lobes. More diffuse fibrosis and loss of alveolar airspace was observed in high-dose BLM-treated segments, while multifocal fibrosis was seen in low-dose BLM-treated segments. Extensive and disorganised collagen deposition occurred in the BLM-treated lobes, compared to controls. Significant loss of lung compliance was also observed in the BLM-treated lobes, which did not occur in controls. Fibrosis comparable to IPF was induced into isolated lung segments, without compromising the respiratory functioning of the animal. This model may have potential for investigating novel therapies for IPF by allowing direct comparison of multiple treatments with internal controls, and sampling and drug delivery that are clinically relevant.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
L-Proline, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
L-Proline, United States Pharmacopeia (USP) Reference Standard
Supelco
L-Proline, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
L-Proline, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
SAFC
L-Proline
Sigma-Aldrich
L-Proline, 99%, FCC, FG
Sigma-Aldrich
L-Proline, ReagentPlus®, ≥99% (HPLC)
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%