Merck
  • Promotion of cell adhesion by low-molecular-weight hydrogel by Lys based amphiphile.
Promotion of cell adhesion by low-molecular-weight hydrogel by Lys based amphiphile.
Materials science & engineering. C, Materials for biological applications (2014-12-11)
Torao Suga, Satoshi Osada, Takayuki Narita, Yushi Oishi, Hiroaki Kodama
ABSTRACT
Hydrogels formed by low-molecular hydrogelators have been used as anti-microbial agents and cell-attachment materials. However the biomedical application of low-molecular gelators is slowly progressing compared to the hydrogels formed by polymer hydrogelator that is applied to biomedical application such as tissue engineering and biomedical regions. To obtain a simple molecular model for potent and prospective usage of low-molecular hydrogelators, we designed a Lys-based hydrogelator which was mimic to the poly cationic poly-l-lysine that promotes cells to attach to a plastic plate nonspecifically. The gel-coating led to cause 10-fold cell attachment compared to no-coating well. Also five-time cells were attached to the well compared to the poly-l-lysine coating. From the competitive assay, these hydrogels could interact with cells through electrostatic interaction between positive charge from -NH3(+) in the hydrogelator and negative charge from substances on the cell surface such as glycosaminoglycans. This strong adhesive ability can be useful for the tissue engineering and molecular glue regions using low-molecular hydrogels in the future.
Sigma-Aldrich
N-Ethyldiisopropylamine solution, suitable for peptide synthesis, ~2 M in 1-methyl-2-pyrrolidinone
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
N-Ethyldiisopropylamine, BASF quality, ≥98.0%
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
HBTU, ≥98.0% (T)
Sigma-Aldrich
N,N-Diisopropylethylamine, purified by redistillation, 99.5%
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Diisopropylethylamine, ReagentPlus®, ≥99%
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
N,N-Diisopropylethylamine, 99.5%, biotech. grade