Merck
  • Alcohol alters the activation of ERK1/2, a functional regulator of binge alcohol drinking in adult C57BL/6J mice.
Alcohol alters the activation of ERK1/2, a functional regulator of binge alcohol drinking in adult C57BL/6J mice.
Alcoholism, clinical and experimental research (2015-02-24)
Abigail E Agoglia, Amanda C Sharko, Kelly E Psilos, Sarah E Holstein, Grant T Reid, Clyde W Hodge
ABSTRACT
Binge alcohol drinking is a particularly risky pattern of alcohol consumption that often precedes alcohol dependence and addiction. The transition from binge alcohol drinking to alcohol addiction likely involves mechanisms of synaptic plasticity and learning in the brain. The mitogen-activated protein kinase (MAPK) signaling cascades have been shown to be involved in learning and memory, as well as the response to drugs of abuse, but their role in binge alcohol drinking remains unclear. The present experiments were designed to determine the effects of acute alcohol on extracellular signaling-related kinases (ERK1/2) expression and activity and to determine whether ERK1/2 activity functionally regulates binge-like alcohol drinking. Adult male C57BL/6J mice were injected with ethanol (EtOH) (3.0 mg/kg, intraperitoneally) 10, 30, or 90 minutes prior to brain tissue collection. Next, mice that were brought to freely consume unsweetened EtOH in a binge-like access procedure were pretreated with the MEK1/2 inhibitor SL327 or the p38 MAPK inhibitor SB239063. Acute EtOH increased pERK1/2 immunoreactivity relative to vehicle in brain regions known to be involved in drug reward and addiction, including the central amygdala and prefrontal cortex. However, EtOH decreased pERK1/2 immunoreactivity relative to vehicle in the nucleus accumbens core. SB239063 pretreatment significantly decreased EtOH consumption only at doses that also produced nonspecific locomotor effects. SL327 pretreatment significantly increased EtOH, but not sucrose, consumption without inducing generalized locomotor effects. These findings indicate that ERK1/2 MAPK signaling regulates binge-like alcohol drinking. As alcohol increased pERK1/2 immunoreactivity relative to vehicle in brain regions known to regulate drug self-administration, SL327 may have blocked this direct pharmacological effect of alcohol and thereby inhibited the termination of binge-like drinking.
Product Description
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
SL327, ≥98% (HPLC), Mixture of E & Z isomers, solid
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethanol, for residue analysis
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Toluidine Blue, for microscopy (Hist., Vit.), 8.74% (ZN (THEORY))
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)