Merck
  • Dispersive micro-solid-phase extraction of benzodiazepines from biological fluids based on polyaniline/magnetic nanoparticles composite.
Dispersive micro-solid-phase extraction of benzodiazepines from biological fluids based on polyaniline/magnetic nanoparticles composite.
Analytica chimica acta (2014-08-31)
Ali Akbar Asgharinezhad, Homeira Ebrahimzadeh, Fatemeh Mirbabaei, Narges Mollazadeh, Nafiseh Shekari
ABSTRACT
In this study, diverse types of Fe3O4 nanocomposites modified by polyaniline, polypyrrole, and aniline-pyrrole copolymer were synthesized through chemical oxidative polymerization process for dispersive-μ-solid phase extraction (D-μ-SPE) in the presence of various dopants. The results showed that the nanocomposite modified by polyaniline with p-toluene sulfonic acid as a dopant demonstrated higher extraction efficiency for lorazepam (LRZ) and nitrazepam (NRZ). Also the synthesized magnetic sorbents were characterized. The nanocomposite sorbent in combination with high performance liquid chromatography-UV detection was applied for the extraction, preconcentration and determination of lorazepam and nitrazepam in urine and plasma samples. Different parameters influencing the extraction efficiency including: sample pH, amount of sorbent, sorption time, elution solvent and its volume, salt content, and elution time were optimized. The obtained optimal conditions were: sample pH, 6; amount of sorbent, 5 mg; sorption time, 5.0 min; elution solvent and its volume, 0.5 mM cethyltrimethyl ammonium bromide in acetonitrile, 150 μL; elution time, 2.0 min and without addition of NaCl. The calibration curves were linear in the concentration range of 1-2000 μg L(-1). The limits of detection (LODs) were achieved in the range of 0.5-1.8 μg L(-1) for NRZ and 0.2-2.0 μg L(-1) for LRZ, respectively. The percent of extraction recoveries and relative standard deviations (n=5) were in the range of 84.0-99.0, 6.1-7.8 for NRZ and 90.0-99.0, 4.1-7.0 for LRZ, respectively. Ultimately, the applicability of the method was successfully confirmed by the extraction and determination of NRZ and LRZ in human urine and plasma samples.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Iron(III) chloride, sublimed grade, ≥99.9% trace metals basis
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Supelco
Aniline, analytical standard
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Iron(III) chloride, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Supelco
Tetrahydrofuran, analytical standard
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Supelco
Methanol, analytical standard
Sigma-Aldrich
1-Propanol, ≥99%, FG
Sigma-Aldrich
Aniline, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Aniline, ReagentPlus®, 99%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Pyrrole, ≥98%, FCC, FG