Merck
  • Combining capillary electrophoresis and next-generation sequencing for aptamer selection.
Combining capillary electrophoresis and next-generation sequencing for aptamer selection.
Analytical and bioanalytical chemistry (2015-01-13)
Kathryn R Riley, Jason Gagliano, Jiajie Xiao, Kara Libby, Shingo Saito, Guo Yu, Roger Cubicciotti, Jed Macosko, Christa L Colyer, Martin Guthold, Keith Bonin
ABSTRACT
Next-generation sequencing (NGS) machines can sequence millions of DNA strands in a single run, such as oligonucleotide (oligo) libraries comprising millions to trillions of discrete oligo sequences. Capillary electrophoresis is an attractive technique to select tight binding oligos or "aptamers" because it requires minimal sample volumes (e.g., 100 nL) and offers a solution-phase selection environment through which enrichment of target-binding oligos can be determined quantitatively. We describe here experiments using capillary transient isotachophoresis (ctITP)-based nonequilibrium capillary electrophoresis of equilibrium mixtures (NECEEM) as a method for selecting aptamers from a randomized library containing a known (29mer) thrombin-binding aptamer. Our capillary electrophoresis (CE)-selected samples were sequenced by the Ion Torrent Personal Genome Machine (PGM) and analyzed for selection efficiency. We show that a single round of CE selection can enrich a randomer synthetic DNA oligo mixture for thrombin-binding activity from 0.4% aptamer content before selection to >15% aptamer content.
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Methylamine solution, 33 wt. % in absolute ethanol ((denatured with 1% toluene))
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Supelco
Trizma® base, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Methylamine solution, 40 wt. % in H2O
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Methylamine solution, 2.0 M in THF
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Methylamine solution, 2.0 M in methanol
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
SAFC
Glycine
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland