Merck
  • In vitro evaluation of effects of sustained anti-TNF release from MPEG-PCL-MPEG and PCL microspheres on human rheumatoid arthritis synoviocytes.
In vitro evaluation of effects of sustained anti-TNF release from MPEG-PCL-MPEG and PCL microspheres on human rheumatoid arthritis synoviocytes.
Journal of biomaterials applications (2014-05-24)
Özge Erdemli, Seza Özen, Dilek Keskin, Ali Usanmaz, Ezgi Deniz Batu, Bülent Atilla, Ayşen Tezcaner
ABSTRACT
Anti-tumor necrosis factor α (TNFα) drugs such as etanercept (ETN) have been mostly used in systemic treatment of rheumatoid arthritis. To eliminate the side effects in long-term treatments and to achieve a local sustained anti-inflammatory effect, a controlled drug delivery system is needed for anti-TNFα drugs. This study aims to develop novel injectable microcarriers of ETN that can provide long-term controlled release of this protein drug upon intra-articular application. In this study, poly(ε-caprolactone) (PCL) and its copolymer with poly(ethylene glycol), methoxypoly(ethylene glycol)-poly(ε-caprolactone)-methoxypoly(ethylene glycol) microspheres (MPEG-PCL-MPEG) were compared for their prospective success in rheumatoid arthritis treatment. Microspheres with smooth surface of a mean particle diameter of approximately 5 μm were prepared with both polymers. MPEG-PCL-MPEG microspheres had higher encapsulation efficiency than PCL microspheres. The activity of encapsulated ETN within MPEG-PCL-MPEG microspheres also retained while 90% of the activity of ETN within PCL microspheres could retain during 90-day release. MPEG-PCL-MPEG microspheres showed faster ETN release compared to PCL microspheres in various release media. Cumulative amounts of ETN released from both types of microspheres were significantly lower in cell culture medium and in synovial fluids than in phosphate buffered saline. This was mainly due to protein adsorption onto microspheres. Hydrophilic MPEG segment enhanced ETN release while preventing protein adsorption on microspheres compared to PCL. Sustained ETN release from microspheres resulted with a significant decrease in pro-inflammatory cytokines (TNFα, IFNγ, IL-6, IL-17) and MMP levels (MMP-3, MMP-13), while conserving viability of fibroblast-like synoviocytes compared to the free drug. Results suggest that MPEG-PCL-MPEG is a potential copolymer of PCL that can be used in development of biomedical materials for effective local treatment purposes in chronic inflammatory arthritis owing to enhanced hydrophilicity. Yet, PCL microspheres are also promising systems having good compatibility to synoviocytes and would be especially the choice for treatment approach requiring longer term and slower release.
Sigma-Aldrich
ε-Caprolactone, 97%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Copper(II) sulfate pentahydrate, 99.995% trace metals basis
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Copper(II) sulfate pentahydrate, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Copper(II) sulfate pentahydrate, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Copper(II) sulfate pentahydrate, 99.999% trace metals basis
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Copper(II) sulfate pentahydrate, ACS reagent, ≥98.0%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Copper(II) sulfate pentahydrate, purum p.a., crystallized, ≥99.0% (RT)
Sigma-Aldrich
Human MMP-3 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Copper(II) sulfate pentahydrate, ReagentPlus®, ≥98.0%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)