Merck
  • Synergistic co-delivery of doxorubicin and paclitaxel by porous PLGA microspheres for pulmonary inhalation treatment.
Synergistic co-delivery of doxorubicin and paclitaxel by porous PLGA microspheres for pulmonary inhalation treatment.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2014-10-12)
Tianshi Feng, Huayu Tian, Caina Xu, Lin Lin, Zhigang Xie, Michael Hon-Wah Lam, Haojun Liang, Xuesi Chen
ABSTRACT
PLGA porous microspheres loaded with doxorubicin (DOX) and paclitaxel (PTX) were developed for in situ treatment of metastatic lung cancer. The synergistic effect of the combined drugs was investigated against B16F10 cells to obtain the optimal prescription for in vivo studies. The combination therapy showed great synergism when DOX was the majority in the combination therapy, while they showed moderate antagonism when PTX is in major. The combination of DOX and PTX at a molar ratio of 5/1 showed the best synergistic therapeutic effect in the free form. However, the drugs exhibited more synergism in the PLGA microspheres at a molar ratio of 2/1, due to the difference in drug release rate. The in vivo study verified the synergism of DOX and PTX at the optimal molar ratio. These results suggested that dual encapsulation of DOX and PTX in porous PLGA microspheres would be a promising technology for long effective lung cancer treatment.
Sigma-Aldrich
Lactic acid solution, ACS reagent, ≥85%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Ammonium bicarbonate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
DL-Lactic acid, ~90% (T)
Sigma-Aldrich
DL-Lactic acid, 85 % (w/w), syrup
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ammonium bicarbonate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
USP
Lactic acid, United States Pharmacopeia (USP) Reference Standard
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Lactic acid, 85%, FCC
Sigma-Aldrich
Glycolic acid solution, technical grade, 70 wt. % in H2O
Sigma-Aldrich
Lactic acid, natural, ≥85%
Sigma-Aldrich
Glycolic acid solution, high purity, 70 wt. % in H2O
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Lactic acid, meets USP testing specifications
Supelco
Glycolic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycolic acid, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Glycolic acid, ReagentPlus®, 99%
USP
Doxorubicin hydrochloride, United States Pharmacopeia (USP) Reference Standard
Supelco
Glycolic Acid, Pharmaceutical Secondary Standard; Certified Reference Material