Merck
  • Red yeast rice repairs kidney damage and reduces inflammatory transcription factors in rat models of hyperlipidemia.
Red yeast rice repairs kidney damage and reduces inflammatory transcription factors in rat models of hyperlipidemia.
Experimental and therapeutic medicine (2014-11-06)
Mei Ding, Daoyuan Si, Wenqi Zhang, Zhaohui Feng, Min He, Ping Yang
ABSTRACT
Xuezhikang (XZK), an extract of red yeast rice, has been widely used for the management of hyperlipidemia and coronary heart disease (CHD); however, the effects of XZK treatment on kidney injury have not yet been fully identified. The aim of the current study was to evaluate the effects of XZK on the kidneys and investigate the related mechanisms in a rat model of hyperlipidemia. Thus, the effect on inflammatory transcription factors and kidney damage was investigated with
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Supelco
3,3′,5,5′-Tetramethylbenzidine, standard for GC
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Supelco
Cholecalciferol (D3), analytical standard
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Sigma-Aldrich
Cholecalciferol, analytical standard
Sigma-Aldrich
Cholecalciferol, meets USP testing specifications
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholecalciferol, ≥98% (HPLC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
4,4′-Diaminodiphenylmethane, ≥97.0% (GC)
Supelco
4,4′-Diaminodiphenylmethane, analytical standard
Cholecalciferol, European Pharmacopoeia (EP) Reference Standard
USP
Cholecalciferol, United States Pharmacopeia (USP) Reference Standard
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Vitamin D3 solution, 100 μg/mL in ethanol, 97% (CP)
Supelco
Vitamin D3 solution, 1 mg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Cholecalciferol (Vitamin D3), Pharmaceutical Secondary Standard; Certified Reference Material