Merck
  • Sulfated polysaccharides isolated from the green seaweed Caulerpa racemosa plays antinociceptive and anti-inflammatory activities in a way dependent on HO-1 pathway activation.
Sulfated polysaccharides isolated from the green seaweed Caulerpa racemosa plays antinociceptive and anti-inflammatory activities in a way dependent on HO-1 pathway activation.
Inflammation research : official journal of the European Histamine Research Society ... [et al.] (2014-03-19)
Natássia Albuquerque Ribeiro, Ticiana Monteiro Abreu, Hellíada Vasconcelos Chaves, Mirna Marques Bezerra, Helena Serra Azul Monteiro, Roberta Jeane Bezerra Jorge, Norma Maria Barros Benevides
ABSTRACT
Marine algae are abundant sources of sulfated polysaccharides with various biological activities. Consequently, their biomolecules are of great of commercial interest. In this study, we investigated the potential antinociceptive activity of a sulfated polysaccharide obtained from the green seaweed Caulerpa racemosa (CrII) and the involvement of the hemoxigenase-1 (HO-1) pathway in its anti-inflammatory effect. We used a systemic evaluation to verify possible toxic effects of Crll after consecutive treatments. Swiss mice and Wistar rats were used for all experiments. In Swiss mice, CrII (0.01, 0.1 and 1.0 mg/kg) significantly reduced the number of abdominal contortions and the duration of paw licking in the second phase after treatment with acetic acid and formalin, respectively. However, CrII was unable to prolong the reaction time of thermally stimulated animals. The anti-inflammatory effect of CrII (0.01, 0.1 and 1.0 mg/kg) was evidenced by a decreased number of leukocytes in the peritoneal cavities of the rats. CrII (0.01, 0.1 and 1.0 mg/kg) also reduced the amount of paw edema induced by carrageenan (Cg) and dextran. The anti-inflammatory effect of CrII was confirmed by reduced levels of myeloperoxidase in the paw tissue of the Cg groups. After inhibition with ZnPP IX, a specific HO-1 phenotype inhibitor, the anti-inflammatory effect of CrII was no longer observed in Cg-induced paw edema tests. Consecutive Crll (1.0 mg/kg) for 14 days did not change any biochemical or histopathological parameters, or cause mortality of mice. CrII did not produce any signs of toxicity and effectively decreased nociception and inflammation. Also, the anti-inflammatory effect of Crll is at least in part dependent on the integrity of the HO-1 pathway.
Sigma-Aldrich
Cetylpyridinium chloride, meets USP testing specifications
Cetylpyridinium chloride, European Pharmacopoeia (EP) Reference Standard
USP
Cetrimonium bromide, United States Pharmacopeia (USP) Reference Standard
USP
Cetylpyridinium chloride, United States Pharmacopeia (USP) Reference Standard
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Sigma-Aldrich
Methylene Blue solution, for microscopy, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Methylene Blue solution, suitable for microbiology
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Chloral hydrate, crystallized, ≥98.0% (T)
Sigma-Aldrich
Methylene Blue solution, 0.05 wt. % in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Methylene blue, certified by the Biological Stain Commission
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Methylene Blue Solution 1.4%(w/v)95%ethanol | 7220-79-3, 1.4 % (w/v) in 95% ethanol
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
Chloral hydrate, ≥99%
Sigma-Aldrich
Methylene Blue solution, for microscopy
SAFC
Hexadecyltrimethylammonium bromide, USP/NF
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis