Merck
  • Ultra performance liquid chromatography-tandem mass spectrometry for the determination of amicarthiazol residues in soil and water samples.
Ultra performance liquid chromatography-tandem mass spectrometry for the determination of amicarthiazol residues in soil and water samples.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-12-03)
Wen-jun Gui, Jie Tian, Chun-xia Tian, Shu-ying Li, You-ning Ma, Guo-nian Zhu
ABSTRACT
A reliable and rapid method has been optimized to determine the residue of amicarthiazol in soil and environmental water samples. After extraction and evaporation, the extraction was carried out with solid phase extraction (SPE) cleanup using HLB cartridge (only soil samples) and for the quantitative determination by ultra performance liquid chromatography-tandem mass spectrometry (UPLC-MS/MS). The resulting residues of amicarthiazol were analyzed by a gradient separation performed on a UPLC system with a C18 column, methanol and water containing 0.1% (v v(-1)) formic acid as the mobile phase in the mode of electrospray positive ionization (ESI(+)) and multiple reaction monitoring (MRM). Results showed that the recoveries for spiked samples were 74.4-97.1% and 72.1-109.9% for soil and water, respectively, with the relative standard deviation (RSD) less than 10.2% when fortified at 10, 100 and 1000μgL(-1). The limits of detection (LODs) and the limits of quantification (LOQs) for matrix matched standards ranged from 0.073-0.425μgL(-1) and 0.243-1.42μgL(-1). The intra-day precision (n=5) and the inter-day precision over 10 days (n=10) for the amicarthiazol in soils and water samples spiked at 100μgL(-1) was 7.9% and 15.9%, respectively. Results indicated that the developed method could be a helpful tool for the controlling and monitoring of the risks posed by amicarthiazol to human health and environment safety.
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%