Merck
  • Direct high-performance liquid chromatographic enantioseparation of secondary amino acids on Cinchona alkaloid-based chiral zwitterionic stationary phases. Unusual temperature behavior.
Direct high-performance liquid chromatographic enantioseparation of secondary amino acids on Cinchona alkaloid-based chiral zwitterionic stationary phases. Unusual temperature behavior.
Journal of chromatography. A (2014-07-20)
István Ilisz, Zsanett Gecse, Zoltán Pataj, Ferenc Fülöp, Géza Tóth, Wolfgang Lindner, Antal Péter
ABSTRACT
Two chiral stationary phases containing a quinine- or a quinidine-based zwitterionic ion-exchanger as chiral selector were applied for the enantioseparation of 27 unusual cyclic secondary α-amino acids. The effects of the nature and concentration of the bulk solvent, the acid and base additives, the structures of the analytes and temperature on the enantioresolution were investigated. To study the effects of temperature and to obtain thermodynamic parameters, experiments were carried out at constant mobile phase compositions in the temperature range -5 to 55 °C. The thermodynamic parameters indicated that in most cases the separations were enthalpy-driven, but some entropy-driven separations were also observed. The sequence of elution of the enantiomers was determined in most cases.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Ammonia, puriss., anhydrous, ≥99.95%
Sigma-Aldrich
trans-4-Hydroxy-L-proline, BioXtra, ≥99.0% (NT)
Supelco
Acetic acid, analytical standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Propylamine, purum, ≥99.0% (GC)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Ethylamine, 97%
Sigma-Aldrich
trans-4-Hydroxy-L-proline, ≥99%
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Propylamine, ≥99%
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Propylamine, 98%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Supelco
Propylamine, analytical standard
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
trans-4-Hydroxy-L-proline, BioReagent, suitable for cell culture, ≥98.5%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
trans-4-Hydroxy-L-proline, analytical standard