Merck
  • Fast determination of harmala alkaloids in edible algae by capillary electrophoresis mass spectrometry.
Fast determination of harmala alkaloids in edible algae by capillary electrophoresis mass spectrometry.
Analytical and bioanalytical chemistry (2015-03-10)
Marcos Tascon, Fernando Benavente, Victoria M Sanz-Nebot, Leonardo G Gagliardi
ABSTRACT
The use of algae as a foodstuff is rapidly expanding worldwide from the East Asian countries, where they are also used for medical care. Harmala alkaloids (HAlk) are a family of bioactive compounds found in the extracts of some plants, including wakame (Undaria pinnatifida), an edible marine invasive algae. HAlks are based on a characteristic β-carboline structure with at least one amino ionizable group. In this work, we report the successful separation of a mixture of six HAlks (harmine, harmaline, harmol, harmalol, harmane, and norharmane) by capillary electrophoresis ion-trap mass spectrometry (CE-IT-MS) in less than 8 min. Optimum separation in fused-silica capillaries and detection sensitivity in positive-ion mode were achieved using a background electrolyte (BGE) with 25 mmol L(-1) ammonium acetate (pH 7.8) and 10% (v/v) methanol, and a sheath liquid with 60:40 (v/v) isopropanol-water and 0.05% (v/v) formic acid. The separation method was validated in terms of linearity, limits of detection and quantification, repeatability, and reproducibility. Later, a sample pretreatment was carefully optimized to determine HAlks in commercial wakame samples with excellent recovery and repeatability. For the complex wakame extracts, the MS-MS fragmentation patterns of the different HAlks were useful to ensure a reliable identification. The complete procedure was validated using the standard-addition calibration method, determining matrix effects on the studied compounds. Harmalol, harmine, and harmaline were naturally present in the samples and were quantified at very low concentrations, ranging from 7 to 24 μg kg(-1) dry algae.
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
Harmine, 98%
Sigma-Aldrich
Harmaline, ≥95%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Supelco
Methanol, analytical standard
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets