Merck
  • Streamlined determination of lysophosphatidylcholines in dried blood spots for newborn screening of X-linked adrenoleukodystrophy.
Streamlined determination of lysophosphatidylcholines in dried blood spots for newborn screening of X-linked adrenoleukodystrophy.
Molecular genetics and metabolism (2014-12-08)
Coleman T Turgeon, Ann B Moser, Lars Mørkrid, Mark J Magera, Dimitar K Gavrilov, Devin Oglesbee, Kimiyo Raymond, Piero Rinaldo, Dietrich Matern, Silvia Tortorelli
ABSTRACT
Pre-symptomatic hematopoietic stem cell transplantation is essential to achieve best possible outcomes for patients with the childhood cerebral form of X-linked adrenoleukodystrophy (X-ALD). We describe a high-throughput method for measurement of C20-C26 lysophosphatidylcholines (LPCs) and biochemical diagnosis of X-ALD using the same dried blood spots (DBS) routinely used for newborn screening. LPCs are extracted from 3-mm DBS punch with methanol containing an isotopically labeled LPC as internal standard. This extract is transferred to a 96-well plate, evaporated and then reconstituted in mobile phase for flow injection analysis tandem mass spectrometry (FIA-MS/MS) in selected reaction monitoring mode for measurement of four different LPCs (C20, C22, C24, C26) and the internal standard (d4-C26-LPC). Analysis time is 1.5min per sample. The mean CVs from the intra- and inter-assay experiments for LPCs were 6.3-15.1% for C20-LPC, 4.4-18.6% for C22-LPC and 4.5-14.3% for C24-LPC. Limits of detection were determined for C20-LPC (LOD=0.03μg/mL), C22-LPC (0.03μg/mL), C24-LPC (0.03μg/mL) and C26-LPC (0.01μg/mL). Reference ranges were established from DBS of 130 newborns and 20 adults. Samples of patients with X-ALD (n=16), peroxisomal biogenesis disorders (n=8), and X-ALD carriers (n=12) were analyzed blindly and all were correctly identified. Analysis of LPC species by FIA-MS/MS is a fast, simple and reliable method to screen for X-ALD and other peroxisomal disorders in DBS. To maximize specificity, abnormal results can be verified by a 2nd tier assay using LC-MS/MS.
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Chloroform, analytical standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%