Merck
  • Comparison of in vitro release rates of acyclovir from cream formulations using vertical diffusion cells.
Comparison of in vitro release rates of acyclovir from cream formulations using vertical diffusion cells.
AAPS PharmSciTech (2014-05-16)
Sumalatha Nallagundla, Srinivas Patnala, Isadore Kanfer
ABSTRACT
Acyclovir, indicated in the treatment of herpes labialis ("cold sores"), is formulated as semisolid topical dosage forms and marketed in numerous countries. Since the formulations of the various acyclovir products may differ from country to country, this study was undertaken to compare the in vitro release of acyclovir from various generic cream products available on the South African and Indian markets using the respective brand/innovator product as the reference product. The in vitro studies were carried out using vertical diffusion cells with a diffusional surface area of 1.767 cm(2) and various commercially available membranes. Normal saline was used as receptor fluid and the temperature maintained at 32 ± 0.5°C. The in vitro release comparisons were based on the recommendations described in the US Food and Drug Administration Draft Guidance for acyclovir ointment and the SUPAC-SS Guidance for non-sterile semisolid dosage forms. The release rates (slope) of the test (T) and the relevant reference product (R) were monitored and compared. The comparative release of acyclovir from the various generic formulations compared with the reference product was found to be within the limits of 75-133.33% with a 90% confidence interval. These experiments indicate that the generic acyclovir cream formulations exhibited release rates that were comparable to the innovator product and could be considered to be bioequivalent.
Product Description
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acycloguanosine, ≥99% (HPLC), powder
USP
Acyclovir, United States Pharmacopeia (USP) Reference Standard
Aciclovir, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Acyclovir, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Millipore
PES Membrane Filter, Strat-M®, filter diam. 25 mm
Millipore
PES Membrane Filter, Strat-M®, filter diam. 47 mm
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%