Merck
  • Lysozyme-triggered epidermal growth factor release from bacterial cellulose membranes controlled by smart nanostructured films.
Lysozyme-triggered epidermal growth factor release from bacterial cellulose membranes controlled by smart nanostructured films.
Journal of pharmaceutical sciences (2014-10-14)
Guilherme Fadel Picheth, Maria Rita Sierakowski, Marco Aurelio Woehl, Lucy Ono, Axel Rulf Cofré, Luana Pasetti Vanin, Roberto Pontarolo, Rilton Alves De Freitas
ABSTRACT
A novel wound-dressing biodevice, sensitive to lysozyme, an enzyme commonly found at infected skin wounds, was assembled by the layer-by-layer deposition of nanopolymeric chitosan and alginate films onto oxidized bacterial cellulose membranes incorporated with epidermal growth factor (EGF). Distinct EGF release profiles were obtained according to specific stimuli caused by infection. In in vitro conditions simulating noninfected wounds, the EGF rate and burst release effect were reduced by three deposited layers (Mt /M∞ of 0.25 at 3 h) in a process dependent on the porosity of the compact chitosan-alginate complex. The importance of the organized structure was revealed when an infected wound was simulated by adding lysozyme to the release medium, thus inducing the formation of a loosely polyelectrolyte architecture that caused rapid EGF diffusion (Mt /M∞ of 0.75 at 30 min). The results indicate that the nanopolymeric layers were capable of slowly releasing EGF as required for normal wound repair and rapidly undergoing architectural transitions that allow the diffusion of massive amounts of drug to enhance the process of re-epithelialization. In summary, the proposed system comprises the roles of both wound dressing and local delivery mechanism to recognize infections and respond with a burst of EGF release.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Sodium bromide, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Supelco
Acetic acid, analytical standard
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Sodium bromide, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Sodium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Sodium bromide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Enrofloxacin for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC