Merck
  • c.620C>T mutation in GATA4 is associated with congenital heart disease in South India.
c.620C>T mutation in GATA4 is associated with congenital heart disease in South India.
BMC medical genetics (2015-05-01)
Saidulu Mattapally, Sheikh Nizamuddin, Kona Samba Murthy, Kumarasamy Thangaraj, Sanjay K Banerjee
ABSTRACT
Congenital heart diseases (CHDs) usually refer to abnormalities in the structure and/or function of the heart that arise before birth. GATA4 plays an important role in embryonic heart development, hence the aim of this study was to find the association of GATA4 mutations with CHD among the south Indian CHD patients. GATA4 gene was sequenced in 100 CHD patients (ASD, VSD, TOF and SV) and 200 controls. Functional significance of the observed GATA4 mutations was analyzed using PolyPhen, SIFT, PMut, Plink, Haploview, ESE finder 3.0 and CONSITE. We observed a total of 19 mutations, of which, one was in 5' UTR, 10 in intronic regions, 3 in coding regions and 5 in 3' UTR. Of the above mutations, one was associated with Atrial Septal Defect (ASD), two were found to be associated with Tetralogy of Fallot (TOF) and three (rs804280, rs4841587 and rs4841588) were strongly associated with Ventricular Septal Defect (VSD). Interestingly, one promoter mutation (-490 to 100 bp) i.e., 620 C>T (rs61277615, p-value = 0.008514), one splice junction mutation (G>A rs73203482; p-value = 9.6e-3, OR = 6.508) and one intronic mutation rs4841587 (p-value = 4.6e-3, OR = 4.758) were the most significant findings of this study. In silico analysis also proves that some of the mutations reported above are pathogenic. The present study found that GATA4 genetic variations are associated with ASD, TOF and VSD in South Indian patients. In silico analysis provides further evidence that some of the observed mutations are pathogenic.
Product Description
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethanol, for residue analysis
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology