Merck
  • Lactococcal 949 group phages recognize a carbohydrate receptor on the host cell surface.
Lactococcal 949 group phages recognize a carbohydrate receptor on the host cell surface.
Applied and environmental microbiology (2015-03-10)
Jennifer Mahony, Walter Randazzo, Horst Neve, Luca Settanni, Douwe van Sinderen
ABSTRACT
Lactococcal bacteriophages represent one of the leading causes of dairy fermentation failure and product inconsistencies. A new member of the lactococcal 949 phage group, named WRP3, was isolated from cheese whey from a Sicilian factory in 2011. The genome sequence of this phage was determined, and it constitutes the largest lactococcal phage genome currently known, at 130,008 bp. Detailed bioinformatic analysis of the genomic region encoding the presumed initiator complex and baseplate of WRP3 has aided in the functional assignment of several open reading frames (ORFs), particularly that for the receptor binding protein required for host recognition. Furthermore, we demonstrate that the 949 phages target cell wall phospho-polysaccharides as their receptors, accounting for the specificity of the interactions of these phages with their lactococcal hosts. Such information may ultimately aid in the identification of strains/strain blends that do not present the necessary saccharidic target for infection by these problematic phages.
Product Description
Supelco
Chloroform, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Pricing and availability is not currently available.
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Pricing and availability is not currently available.
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, natural, 97%, FG
Pricing and availability is not currently available.
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Pricing and availability is not currently available.
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Pricing and availability is not currently available.
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Pricing and availability is not currently available.
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Pricing and availability is not currently available.
Supelco
Phenol, PESTANAL®, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Pricing and availability is not currently available.