Merck
  • Semiconductor polymer dots induce proliferation in human gastric mucosal and adenocarcinoma cells.
Semiconductor polymer dots induce proliferation in human gastric mucosal and adenocarcinoma cells.
Macromolecular bioscience (2014-12-17)
Shouying Li, Jia Chen, Geng Chen, Qiong Li, Kai Sun, Zhen Yuan, Weiping Qin, Hong Xu, Changfeng Wu
ABSTRACT
We investigated the cellular uptake behavior and cell viability of semiconducting polymer dots (Pdots) on human gastric adenocarcinoma (SGC-7901) cells and human gastric mucosal (GES-1) cells. MTT studies indicate the Pdot treatment induces obvious cell proliferation in both types of cell lines. We performed further investigations such as reactive oxygen species (ROS) generation and mitochondrial membrane potential (MMP) change, which indicate that the cell proliferation is in good agreement with the increase in the ROS and MMP levels. Moreover, expression of protein kinase B (Akt) decreased as the Pdot concentration increases, but the expression of protein dual-phosphorylated Erk (p-Erk) and phosphorylated c-Jun N-terminal kinases (p-JNK) were increased. These effects indicated that the Pdots could promote the growth of SGC-7901 cells and GES-1 cells by appropriately regulating the expressions of protein Akt, p-Erk, and p-JNK.
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Maleic anhydride, 99%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Styrene, ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%
Supelco
Styrene, analytical standard
Sigma-Aldrich
Maleic anhydride, puriss., ≥99.0% (NT)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri