Merck
Caffeine inhibits hepatitis C virus replication in vitro.
Archives of virology (2014-12-11)
Mariana N Batista, Bruno M Carneiro, Ana Cláudia S Braga, Paula Rahal
ABSTRACT
Hepatitis C is considered the major cause of cirrhosis and hepatocellular carcinoma. Conventional treatment is not effective against some hepatitis C virus (HCV) genotypes; therefore, new treatments are needed. Coffee and, more recently, caffeine, have been found to have a beneficial effect in several disorders of the liver, including those manifesting abnormal liver biochemistry, cirrhosis and hepatocellular carcinoma. Caffeine acts directly by delaying fibrosis, thereby improving the function of liver cellular pathways and interfering with pathways used by the HCV replication cycle. In the current study, the direct relationship between caffeine and viral replication was evaluated. The Huh-7.5 cell line was used for transient infections with FL-J6/JFH-5'C19Rluc2AUbi and to establish a cell line stably expressing SGR-Feo JFH-1. Caffeine efficiently inhibited HCV replication in a dose-dependent manner at non-cytotoxic concentrations and demonstrated an IC50 value of 0.7263 mM after 48 h of incubation. These data demonstrate that caffeine may be an important new agent for anti-HCV therapies due to its efficient inhibition of HCV replication at non-toxic concentrations.
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Cyclosporin A, 97.0-101.5% (on dried basis)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Cyclosporin A, BioReagent, from Tolypocladium inflatum, for molecular biology, ≥95%
Sigma-Aldrich
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Cyclosporin A, VETRANAL®, analytical standard
Supelco
Melting point standard 235-237°C, analytical standard
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
SAFC
HEPES
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
SAFC
HEPES
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material