Merck
  • Transcriptome sequencing and metabolite analysis reveals the role of delphinidin metabolism in flower colour in grape hyacinth.
Transcriptome sequencing and metabolite analysis reveals the role of delphinidin metabolism in flower colour in grape hyacinth.
Journal of experimental botany (2014-05-03)
Qian Lou, Yali Liu, Yinyan Qi, Shuzhen Jiao, Feifei Tian, Ling Jiang, Yuejin Wang
ABSTRACT
Grape hyacinth (Muscari) is an important ornamental bulbous plant with an extraordinary blue colour. Muscari armeniacum, whose flowers can be naturally white, provides an opportunity to unravel the complex metabolic networks underlying certain biochemical traits, especially colour. A blue flower cDNA library of M. armeniacum and a white flower library of M. armeniacum f. album were used for transcriptome sequencing. A total of 89 926 uni-transcripts were isolated, 143 of which could be identified as putative homologues of colour-related genes in other species. Based on a comprehensive analysis relating colour compounds to gene expression profiles, the mechanism of colour biosynthesis was studied in M. armeniacum. Furthermore, a new hypothesis explaining the lack of colour phenotype of the grape hyacinth flower is proposed. Alteration of the substrate competition between flavonol synthase (FLS) and dihydroflavonol 4-reductase (DFR) may lead to elimination of blue pigmentation while the multishunt from the limited flux in the cyanidin (Cy) synthesis pathway seems to be the most likely reason for the colour change in the white flowers of M. armeniacum. Moreover, mass sequence data obtained by the deep sequencing of M. armeniacum and its white variant provided a platform for future function and molecular biological research on M. armeniacum.
Product Description
Supelco
(+)-Catechin, analytical standard
Supelco
Delphinidin chloride, analytical standard
Supelco
Luteolin, analytical standard
Sigma-Aldrich
Luteolin, ≥98% (TLC), powder
Luteolin, primary reference standard
Sigma-Aldrich
(−)-Epicatechin, ≥90% (HPLC)
Sigma-Aldrich
(±)-Naringenin, ≥95%
Sigma-Aldrich
(−)-Epicatechin, ≥98% (HPLC), from green tea
Supelco
(±)-Naringenin, analytical standard
Sigma-Aldrich
Naringenin, natural (US), 98%
Supelco
Quercetin, Pharmaceutical Secondary Standard; Certified Reference Material
Epicatechin, primary reference standard
USP
Quercetin, United States Pharmacopeia (USP) Reference Standard
Supelco
(−)-Epicatechin, analytical standard
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid