Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sign Into View Organizational & Contract Pricing Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur™, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium Fluoride Solution
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sign Into View Organizational & Contract Pricing Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Anti-REST Antibody, Upstate®, from rabbit
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri™
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sign Into View Organizational & Contract Pricing Supelco
2-Propanol, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
Fluoride ion solution for ISE, 0.1 M F-, analytical standard (for ion-selective electrodes)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sign Into View Organizational & Contract Pricing