Merck
  • Chemometric strategy for untargeted lipidomics: biomarker detection and identification in stressed human placental cells.
Chemometric strategy for untargeted lipidomics: biomarker detection and identification in stressed human placental cells.
Analytica chimica acta (2014-12-07)
Eva Gorrochategui, Josefina Casas, Cinta Porte, Sílvia Lacorte, Romà Tauler
ABSTRACT
A lipidomic study was developed in a human placental choriocarcinoma cell line (JEG-3) exposed to tributyltin (TBT) and to a mixture of perfluorinated chemicals (PFCs). The method was based on the application of multivariate curve resolution-alternating least squares (MCR-ALS) to data sets obtained by ultra-high performance liquid chromatography coupled to time-of-flight mass spectrometry (UHPLC-TOF-MS) using an untargeted approach. Lipids from exposed JEG-3 cells were solid-liquid extracted and analyzed by UHPLC-TOF-MS in full scan mode, together with control samples. Raw UHPLC-TOF-MS data of the different cell samples were subdivided into 20 distinct chromatographic windows and each window was further organized in a column-wise augmented data matrix, where data from every sample was in an individual data matrix. Then, the 20 new augmented data matrices were modeled by MCR-ALS. A total number of 86 components were resolved and a statistical comparative study of their elution profiles showed distinct responses for the lipids of exposed versus control cells, evidencing a lipidome disruption attributed to the presence of the xenobiotics. Results from one-way ANOVA followed by a multiple comparisons test and from discriminant partial least squares (PLS-DA) analysis were compared as usual strategies for the determination of potential biomarkers. Identification of 24 out of the 33 proposed biomarkers contributed to the better understanding of the effects of PFCs and TBT in the lipidome of human placental cells. Overall, this study proposes an innovative untargeted LC-MS MCR-ALS approach valid for -omic sciences such as lipidomics.
Product Description
Supelco
Heptadecafluorooctanesulfonic acid solution, 100 μg/mL in methanol, analytical standard
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Butylated Hydroxytoluene, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Butylated hydroxytoluene, United States Pharmacopeia (USP) Reference Standard
Supelco
Chloroform, analytical standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Supelco
Heptafluorobutyric acid solution, 0.5 M in H2O, LiChropur, suitable for ion pair chromatography
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Undecafluorohexanoic acid, ≥97.0% (T)
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Perfluorooctanoic acid, 95%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%