Merck
  • Multi-responsive polymer micelles as ellipticine delivery carriers for cancer therapy.
Multi-responsive polymer micelles as ellipticine delivery carriers for cancer therapy.
Anticancer research (2015-02-11)
Martin Studenovský, Ondřej Sedláček, Martin Hrubý, Jiří Pánek, Karel Ulbrich
ABSTRACT
In the present study, we describe the synthesis and physicochemical properties of a novel pH- and thermoresponsive micellar drug delivery system for an anticancer ellipticinium derivative based on the triblock copolymer poly(ethylene oxide)-block-[tert-butylacrylamide-co-6-(N-methacryloylamino)hexanoic acid hydrazide]-block-poly(ethylene oxide). The system was designed to meet the basic criteria required for drug carrier systems, namely, solubility in water (overcoming the insolubility of ellipticine), satisfactory drug loading, particle size suitable for an efficient enhanced permeability and retention effect and adequate stability in blood plasma (pH 7.4) followed by rapid drug release in tumors or tumor cell endosomes (pH<6.5). The copolymer in the form of a unimer can be eliminated by kidneys because the weight-average molecular weight of 21 kDa is sufficiently below the renal threshold. The half-life of drug release in a pH 5.0 buffer solution (pH of a late endosome) was ~45 h, but a negligible amount of the free ellipticine derivative was detected at pH 7.4 (pH of blood). Consequently, this supramolecular polymer conjugate is a good candidate for the delivery of ellipticine-based drugs and will therefore be subjected to more detailed studies.
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
N,N-Dimethylformamide, analytical standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), 98%
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), purum, ≥98.0% (GC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Methacryloyl chloride, 97%, contains ~200 ppm monomethyl ether hydroquinone as stabilizer
Supelco
Acetonitrile, analytical standard