Merck
  • Selective Interference Targeting of Lnk in Umbilical Cord-Derived Late Endothelial Progenitor Cells Improves Vascular Repair, Following Hind Limb Ischemic Injury, via Regulation of JAK2/STAT3 Signaling.
Selective Interference Targeting of Lnk in Umbilical Cord-Derived Late Endothelial Progenitor Cells Improves Vascular Repair, Following Hind Limb Ischemic Injury, via Regulation of JAK2/STAT3 Signaling.
Stem cells (Dayton, Ohio) (2014-12-30)
Sang Hun Lee, Kyeung Bin Lee, Jun Hee Lee, Songhwa Kang, Hwi Gon Kim, Takayuki Asahara, Sang Mo Kwon
ABSTRACT
The Lnk adaptor protein is a strong negative regulator that affects self-renewal of hematopoietic stem cells and vascular repair in injured tissues. However, the signaling mechanisms through which these proteins influence the vascular regeneration function of endothelial progenitor cells (EPCs) remain unknown. In this study, we investigated the effect of Lnk-targeted small interfering RNA (si-lnk) on the clonogenic proliferative potential and vascular regenerative function of EPCs and the activation of the JAK/STAT3 signaling pathway. Treatment with stem cell factor (SCF) increased the clonogenic proliferation of si-lnk EPCs. Importantly, activation of the JAK2/STAT3 pathway was enhanced in SCF-sensitized si-lnk EPCs. In a hind limb model of ischemia, transplantation of si-lnk EPCs increased the blood flow ratio, capillary density, proliferation, and survival of transplanted cells, and the secretion of pivotal angiogenic cytokines at ischemic sites. These results provide strong evidence that si-lnk regulates the clonogenic proliferative potential of EPCs through the activation of the JAK2/STAT3 signaling pathway, thereby accelerating angiogenesis and promoting repair in injured hind limb ischemia. Stem Cells 2014;33:1490-1500.
Sigma-Aldrich
Vinylene carbonate, contains ≤2% BHT as stabilizer, 97%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Stat3
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Sh2b3
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
MISSION® esiRNA, targeting human SH2B3
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
MISSION® esiRNA, targeting human STAT3