Merck
  • Copy-neutral loss of heterozygosity is prevalent and a late event in the pathogenesis of FLT3/ITD AML.
Copy-neutral loss of heterozygosity is prevalent and a late event in the pathogenesis of FLT3/ITD AML.
Blood cancer journal (2014-05-03)
D L Stirewalt, E L Pogosova-Agadjanyan, K Tsuchiya, J Joaquin, S Meshinchi
ABSTRACT
Patients with high FLT3 internal tandem duplication allelic ratios (FLT3/ITD-ARs) have a poor prognosis. Single-nucleotide polymorphism/comparative genomic hybridization, single-cell PCR and colony-forming assays were used to evaluate genotypic evolution of high FLT3/ITD-ARs in 85 acute myeloid leukemia (AML) patients. Microarrays were used to examine molecular pathways disrupted in leukemic blasts with high FLT3/ITD-ARs. Copy-neutral loss of heterozygosity (CN-LOH) was identified at the FLT3 locus in diagnostic samples with high FLT3/ITD-ARs (N=11), but not in samples with low FLT3/ITD-ARs (N=24), FLT3-activating loop mutations (N=11) or wild-type FLT3 (N=39). Single-cell assays showed that homozygous FLT3/ITD genotype was present in subsets of leukemic blasts at diagnosis but became the dominant clone at relapse. Less differentiated CD34(+)/CD33(-) progenitor colonies were heterozygous for FLT3/ITD, whereas more differentiated CD34(+)/CD33(+) progenitor colonies were homozygous for FLT3/ITD. Expression profiling revealed that samples harboring high FLT3/ITD-ARs aberrantly expressed genes within the recombination/DNA repair pathway. Thus, the development of CN-LOH at the FLT3 locus, which results in high FLT3/ITD-ARs, likely represents a late genomic event that occurs after the acquisition of the FLT3/ITD. Although the etiology underlying the development of CN-LOH remains to be clarified, the disruption in recombination/DNA repair pathway, which is present before the development of LOH, may have a role.
Sigma-Aldrich
FLT3 (571-993), active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide solution